Skip to Content
Merck
All Photos(1)

Key Documents

86717

Sigma-Aldrich

Tetraallylsilane

≥97.0% (GC)

Synonym(s):

Tetra(2-propenyl)silane

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$241.00

$241.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$241.00

About This Item

Linear Formula:
(H2C=CHCH2)4Si
CAS Number:
Molecular Weight:
192.37
Beilstein:
1750943
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$241.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.485 (lit.)
n20/D 1.485

bp

90 °C/3 mmHg (lit.)

density

0.831 g/mL at 25 °C (lit.)

SMILES string

C=CC[Si](CC=C)(CC=C)CC=C

InChI

1S/C12H20Si/c1-5-9-13(10-6-2,11-7-3)12-8-4/h5-8H,1-4,9-12H2

InChI key

AKRQMTFHUVDMIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service