Skip to Content
Merck
All Photos(1)

Key Documents

90921

Sigma-Aldrich

2-Acetamido-1,2-dideoxynojirimycin

≥98.0% (TLC)

Synonym(s):

2-Acetamido-1,2,5-trideoxy-1,5-imino-D-glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N2O4
CAS Number:
Molecular Weight:
204.22
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98.0% (TLC)

form

powder

storage temp.

2-8°C

SMILES string

CC(=O)N[C@H]1CN[C@H](CO)[C@@H](O)[C@@H]1O

InChI

1S/C8H16N2O4/c1-4(12)10-5-2-9-6(3-11)8(14)7(5)13/h5-9,11,13-14H,2-3H2,1H3,(H,10,12)/t5-,6+,7+,8+/m0/s1

InChI key

GBRAQQUMMCVTAV-LXGUWJNJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Acetamido-1,2-dideoxynojirimycin (2-ADN) is used as an inhibitor (transition analogue) to identify, purify, differentiate and characterized N-acetylglucosaminidase(s) (GlcNAcase). 2-Acetamido-1,2-dideoxynojirimycin is used as a ligand for the affinity purification of N-acetylglucosaminidases.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yeon Kyu Kim et al.
Glycobiology, 19(3), 301-308 (2008-12-05)
Most insect cells have a simple N-glycosylation process and consequently paucimannosidic or simple core glycans predominate. Previously, we have shown that paucimannosidic N-glycan structures are dominant in Drosophila S2 cells. It has been proposed that beta-N-acetylglucosaminidase (GlcNAcase), a hexosaminidase in
B Woynarowska et al.
Anticancer research, 12(1), 161-166 (1992-01-01)
Human ovarian carcinoma (HOC) cell beta-N-acetylglucosaminidase (beta-NAG, EC 3.2.1.30) was found to be present in three isoenzymatic forms. All three forms were capable of degrading ECM. Therefore, inhibitors of beta-NAG were sought as potential anti-invasive agents. Two sugar analogs, 2-acetamido-2-deoxy-1,5-gluconolactone
H Böshagen et al.
Carbohydrate research, 164, 141-148 (1987-07-01)
The synthesis of 2-acetamido-1,2-dideoxynojirimycin (2-acetamido-1,2,5-tri-deoxy-1,5-imino-D-glucitol) by a double inversion procedure starting from 1-deoxynojirimycin is reported. The key intermediates were the selectively protected N-benzyl-1,5-dideoxy-1,5-imino-4,6-O-isopropylidene-D-mannitol, the triflate ester N-benzyl-3-O-benzyl-1,5-dideoxy-1,5-imino-4,6-O-isopropylidene-2-O- (tri-fluoromethylsulfonyl)-D-mannitol, and 2-azido-N-benzyl-3-O-benzyl-1,2,5-tri-deoxy-1,5-imino-4,6-O- isopropylidene-D-glucitol, readily obtained in a sequence from 1-deoxynojirimycin. Thus 1-deoxynojirimycin
G Gradnig et al.
Carbohydrate research, 287(1), 49-57 (1996-06-07)
6-Azido-1,3,4-tri-O-benzyl-6-deoxy-D-fructofuranose can be easily obtained in two steps from the known 6,6'-diazido-6,6'-dideoxysucrose (available in two steps from sucrose) and cyclized by controlled hydrogenation and concomitant intramolecular reductive amination to give 3,4,6-tri-O-benzyl-1,5-dideoxy-1,5-imino-D-mannitol, a partially protected derivative of 1-deoxymannojirimycin. After N-protection, position
K W Zhao et al.
Protein expression and purification, 19(1), 202-211 (2000-06-02)
alpha-N-Acetylglucosaminidase (EC 3.2.1.50) is a lysosomal enzyme that is deficient in the genetic disorder Sanfilippo syndrome type B. To study the human enzyme, we expressed its cDNA in Lec1 mutant Chinese hamster ovary (CHO) cells, which do not synthesize complex

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service