Skip to Content
Merck
All Photos(1)

Key Documents

85775

Sigma-Aldrich

Stearyl stearate

≥98.0% (GC)

Synonym(s):

Stearic acid stearyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$294.00
5 G
$1,370.00

$294.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$294.00
5 G
$1,370.00

About This Item

Linear Formula:
CH3(CH2)16COO(CH2)17CH3
CAS Number:
Molecular Weight:
536.96
Beilstein:
1807499
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

$294.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

synthetic

Assay

≥98.0% (GC)

form

powder

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C36H72O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-38-36(37)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-35H2,1-2H3

InChI key

NKBWPOSQERPBFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dorothy M Duffy et al.
Langmuir : the ACS journal of surfaces and colloids, 20(18), 7630-7636 (2004-08-25)
Living organisms can control the size, shape, and structure of minerals. Attempts to reproduce this biological control in the laboratory often use Langmuir monolayers of long-chain carboxylic acids. We use large-scale molecular dynamics simulations to calculate the interfacial energies of
K Maeda et al.
Journal of chromatography, 221(2), 199-204 (1980-12-12)
A gas chromatographic--mass spectrometric analysis was used to separate and identify abnormal compounds in the nail of psoriatic patients. The nail was extracted with heated ethanol, and the extract was analyzed with and without trimethylsilylation. Tetradecanoic acid octadecyl ester, hexadecanoic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service