Skip to Content
Merck
All Photos(1)

Key Documents

248061

Sigma-Aldrich

Octafluoronaphthalene

96%

Synonym(s):

1,2,3,4,5,6,7,8-Octafluoronaphthalene, Perfluoronaphthalene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$63.00

$63.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$63.00

About This Item

Empirical Formula (Hill Notation):
C10F8
CAS Number:
Molecular Weight:
272.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$63.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

solid

mp

87-88 °C (lit.)

functional group

fluoro

SMILES string

Fc1c(F)c(F)c2c(F)c(F)c(F)c(F)c2c1F

InChI

1S/C10F8/c11-3-1-2(5(13)9(17)7(3)15)6(14)10(18)8(16)4(1)12

InChI key

JDCMOHAFGDQQJX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Collings et al.
Acta crystallographica. Section C, Crystal structure communications, 57(Pt 7), 870-872 (2001-07-10)
The structure of the title complex, C10F8*C14H10, comprises mixed stacks of alternating diphenylacetylene and octafluoronaphthalene molecules, both lying at inversion centres and parallel to within 8.6 (1) degrees, in contrast with the herring-bone packing observed in crystals of either pure
Qimeng Jiang et al.
Polymers, 12(7) (2020-07-28)
Solid acid catalysts generally show the disadvantage of low acid amount and low recycling rate. To solve these problems, corn stalk-based solid acid catalysts were synthesized through carbonization and sulfonation processes in this work. The results showed that besides the
A phase transformation with no change in space group symmetry; octafluoronaphthalene.
Pawley GS and Dietrich O-W.
Journal of Physics C: Solid State Physics, 8(16), 2549-2549 (1975)
Takanori Iwasaki et al.
Chemistry, an Asian journal, 15(8), 1349-1354 (2020-02-28)
Changes in the photophysical properties of pyrene (Py)-octafluoronaphthalene (OFN) co-crystals (Py⋅OFN) upon mechanical stimuli are described herein. The Py⋅OFN co-crystal showed a mechano-induced bathochromic shift in emission, and a similar tendency was observed for the 1,3,6,8-tetramethylpyrene-OFN co-crystal. These shifts are
Polycyclic fluoroaromatic compounds. Part XII.[1] The extent of 1-substitution in octafluoronaphthalene.
Burdon J and Rimmington TW.
Journal of Fluorine Chemistry, 27(3), 257-261 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service