811-500
Human Preadipocyte Growth Medium (500ml)
Synonym(s):
Primary Cell Culture Media
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
Quality Level
shelf life
6 mo.
manufacturer/tradename
Cell Applications, Inc
shipped in
wet ice
storage temp.
2-8°C
General description
All-in-one ready-to-use media
Application
Human Preadipocyte Growth Medium (500ml) has been used in culturing of human preadipocytes.
Quality
Each lot was tested for the ability to support plating, spreading and proliferation
Preparation Note
contained in protocol
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Lab on a chip, 19(2), 241-253 (2018-12-20)
Infiltration of immune cells into adipose tissue is associated with chronic low-grade inflammation in obese individuals. To better understand the crosstalk between immune cells and adipocytes, in vivo-like in vitro models are required. Conventionally transwell culture plates are used for
Biomedical microdevices, 19(3), 18-18 (2017-03-31)
Adipocytes have gained significant attention recently, because they are not only functioning as energy storage but also as endocrine cells. Adipocytes secret various signaling molecules, including adiponectin, MCP-1, and IL-6, termed collectively as "adipokines". Adipokines regulate glucose metabolism, thereby play
Molecular and cellular endocrinology, 506, 110744-110744 (2020-02-07)
Triiodothyronine (T3) and irisin (I) can modulate metabolic status, increase heat production, and promote differentiation of white adipose tissue (WAT) into brown adipose tissue (BAT). Herein, human subcutaneous white adipocytes were treated with 10 nM T3 or 20 nM I for 24 h
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service