77237
Percoll®
colloidal silica coated with polyvinylpyrrolidone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
Recommended Products
storage temp.
2-8°C
Application
Percoll consists of colloidal silica particles of 15-30 nm diameter (23% w/w in water) which have been coated with polyvinylpyrrolidone (PVP). It is used in cell separation and organelle isolation.
Percoll consists of colloidal silica particles of 15-30 nm diameter (23% w/w in water) which have been coated with polyvinylpyrrolidone (PVP). It is used to establish non-toxic low viscosity, low osmolarity density gradients that can be used to isolate cells, organelles and viruses. It has been used to separate inner and outer membrane vesicles from E. coli. Percoll is isosmotic throughout the gradient, non-toxic, and is easily removed from the purified materials.
Other Notes
A unique density gradient medium designed to separate cells, subcellular particles and larger viruses under mild conditions. Review; Separation of inner and outer membrane vesicles from E.coli
Legal Information
Percoll is a registered trademark of Cytiva
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S Morein et al.
Analytical biochemistry, 216(1), 47-51 (1994-01-01)
A rapid and simple method for separating and isolating the inner and outer membranes of Escherichia coli is described. Membrane vesicles were prepared either by passing the bacteria through a French press or by conversion of the cells to spheroplasts
Density gradients prepared from colloidal silica particles coated by polyvinylpyrrolidone (Percoll).
H Pertoft et al.
Analytical biochemistry, 88(1), 271-282 (1978-07-15)
Jieli Chen et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 29(5), 911-920 (2009-02-19)
We tested the hypothesis that Niaspan (a prolonged release formulation of niacin) increases tumor necrosis factor-alpha-converting enzyme (TACE) expression and Notch signaling activity and promotes arteriogenesis after stroke. Rats were subjected to middle cerebral artery occlusion and were treated with
Yanping P Chen et al.
The Journal of eukaryotic microbiology, 56(2), 142-147 (2009-05-22)
Nosema ceranae, a microsporidian parasite originally described from Apis cerana, has been found to infect Apis melllifera and is highly pathogenic to its new host. In the present study, data on the ultrastructure of N. ceranae, presence of N. ceranae-specific
L Tarsa et al.
Neuropeptides, 43(1), 47-52 (2008-11-21)
The role of neuronal growth factors in synaptic maturation of sensory neurons, including trigeminal ganglion (TG) neurons, remains poorly understood. Here, we show that nerve growth factor (NGF) regulates the intracellular distribution of the synaptic vesicle protein synaptophysin (Syp) in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service