Skip to Content
Merck
All Photos(1)

Key Documents

72335

Sigma-Aldrich

9-(2,2-Dicyanovinyl)julolidine

BioReagent, suitable for fluorescence, ≥97.0% (HPLC)

Synonym(s):

9-Julolidinylmethylenemalononitrile, DCVJ

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15N3
CAS Number:
Molecular Weight:
249.31
Beilstein:
413882
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Quality Level

Assay

≥97.0% (HPLC)

form

crystals

solubility

DMSO: soluble
chloroform: soluble

fluorescence

λex 433 nm; λem 500 nm in 50 mM Tris pH 8.0; 40% glycerol

suitability

suitable for fluorescence

SMILES string

N#C\C(=C/c1cc2CCCN3CCCc(c1)c23)C#N

InChI

1S/C16H15N3/c17-10-13(11-18)7-12-8-14-3-1-5-19-6-2-4-15(9-12)16(14)19/h7-9H,1-6H2

InChI key

LROAUBRDKLVBCP-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Fluorescent probe for binding to proteins etc., quantum yield increases by decreasing free rotation; abs at ~450 nm, em at 480-505 nm (solvent dep.)

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Haidekker et al.
American journal of physiology. Heart and circulatory physiology, 278(4), H1401-H1406 (2000-04-06)
Fluid shear stress (FSS) has been shown to be an ubiquitous stimulator of mammalian cell metabolism. Although many of the intracellular signal transduction pathways have been characterized, the primary mechanoreceptor for FSS remains unknown. One hypothesis is that the cytoplasmic
Andrea Hawe et al.
Pharmaceutical research, 27(2), 314-326 (2009-12-31)
The aim was to evaluate fluorescent molecular rotors (DCVJ and CCVJ), which are mainly sensitive to viscosity, for the characterization of polysorbate-containing IgG formulations and compare them to the polarity-sensitive dyes ANS, Bis-ANS and Nile Red. IgG formulations with polysorbate
A Bhattacharyya et al.
Indian journal of biochemistry & biophysics, 32(6), 442-446 (1995-12-01)
Di(cyano vinyl)julolidine (DCVJ) is a fluorescent probe which has been used to monitor the local mobility of its binding sites on proteins. It shows a concentration dependence of its emission spectrum in water. At higher DCVJ concentrations, a longer wavelength
A Bhattacharya et al.
Protein science : a publication of the Protein Society, 5(10), 2029-2036 (1996-10-01)
9-(Dicyanovinyl) julolidine (DCVJ) is a fluorescent probe, which binds to a unique site on the tubulin dimer and exhibits different properties that are dependent upon its oligomeric state (Kung & Reed, 1989). DCVJ binds to tubulin, the tubulin-colchicine complex, and
C E Kung et al.
Biochemistry, 28(16), 6678-6686 (1989-08-08)
9-(Dicyanovinyl)julolidine (DCVJ) is a fluorescent dye whose intramolecular rotational relaxation is solvent dependent. Since its quantum yield increases with decreasing free volume, this molecule has been very useful in monitoring synthetic polymer reactions and measuring local microviscosity changes in phospholipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service