Skip to Content
Merck
All Photos(1)

Key Documents

70550

Sigma-Aldrich

Naphthol Green B

for microscopy (Hist.), for complexometry

Synonym(s):

Acid Green 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$96.50
100 G
$271.00
500 G
$1,140.00

$96.50


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$96.50
100 G
$271.00
500 G
$1,140.00

About This Item

Empirical Formula (Hill Notation):
C30H15FeN3Na3O15S3
CAS Number:
Molecular Weight:
878.46
Colour Index Number:
10020
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$96.50


Estimated to ship onMay 28, 2025


Request a Bulk Order

grade

for complexometry
for microscopy (Hist.)

form

powder

technique(s)

titration: suitable

solubility

methanol: 0.01 g/10 mL

λmax

714 nm

εmax

≥80 at 709-719 nm in water

suitability

suitable for microscopy
suitable for suitability for complexometry

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[Na+].[O-]S(=O)(=O)c1ccc2c(N=O)c(O[Fe](Oc3ccc4cc(ccc4c3N=O)S([O-])(=O)=O)Oc5ccc6cc(ccc6c5N=O)S([O-])(=O)=O)ccc2c1

InChI

1S/3C10H7NO5S.Fe.3Na/c3*12-9-4-1-6-5-7(17(14,15)16)2-3-8(6)10(9)11-13;;;;/h3*1-5,12H,(H,14,15,16);;;;/q;;;+3;3*+1/p-6

InChI key

JMXROTHPANUTOJ-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service