Skip to Content
Merck
All Photos(1)

Key Documents

68616

Sigma-Aldrich

Atto 594 Protein Labeling Kit

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,910.00

$1,910.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
1 KIT
$1,910.00

About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

$1,910.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

product line

BioReagent

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 594 nm; λem 625 nm in 0.1 M phosphate buffer, pH 7.0 (recommended)

suitability

suitable for fluorescence

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This kit contains sufficient amounts of reactive dye, buffers and protein purification sets for performing five labeling reactions (1 mg protein each) and for the subsequent purification of the labeled protein.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Protein labeling kits with Atto and Tracy dyes provide easy fluorescent labeling of purified proteins, enzymes, and antibodies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service