Skip to Content
Merck

Skip To

51146

Atto 550 Protein Labeling Kit

BioReagent, suitable for fluorescence

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Check Cart for Availability
$2,070.00

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Storage temp.:
2-8°C

$2,070.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 544 nm; λem 576 nm in 0.1 M phosphate buffer, pH 7.0 (recommended)

suitability

suitable for fluorescence

storage temp.

2-8°C

General description

This kit contains sufficient amounts of reactive dye, buffers and protein purification sets for performing five labeling reactions (1 mg protein each) and for the subsequent purification of the labeled protein.

Application

Atto fluorescent labels are designed for high sensitivity applications, including single molecule detection. Atto labels have rigid structures that do not show any cis-trans-isomerization. Thus these labels display exceptional intensity with minimal spectral shift on conjugation. Atto 550, which is similar to Cy3, may be useful in applications such as fluorescence resonance energy transfer (FRET) and as a tag for molecules such as secondary antibodies. Atto 550 Protein Labeling Kit contains Atto 550 succinimidyl ester which enables conjugation of Atto 550 to the primary amino groups of the proteins and other molecules.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
686167650838371
fluorescence

λex 544 nm; λem 576 nm in 0.1 M phosphate buffer, pH 7.0 (recommended)

fluorescence

λex 594 nm; λem 625 nm in 0.1 M phosphate buffer, pH 7.0 (recommended)

fluorescence

λex 647 nm; λem 661 nm in 0.1 M phosphate buffer, pH 7.0 (recommended)

fluorescence

λex 488 nm; λem 520 nm in 0.1 M phosphate buffer, pH 7.0 (recommended)

manufacturer/tradename

ATTO-TEC GmbH

manufacturer/tradename

ATTO-TEC GmbH

manufacturer/tradename

ATTO-TEC GmbH

manufacturer/tradename

ATTO-TEC GmbH

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

suitable for fluorescence

product line

BioReagent

product line

BioReagent

product line

BioReagent

product line

BioReagent


Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters