Skip to Content
Merck
All Photos(1)

Documents

59321C

SAFC

Dulbecco′s Phosphate Buffered Saline

liquid, sterile-filtered, DPBS Modified, without calcium, without magnesium, suitable for cell culture

Synonym(s):

DPBS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Application

Dulbecco′s Phosphate Buffered Saline has been used to resuspend phosphatidic acid (PA) or phosphoserine (PS). It has also been used for cell loading with Ca2+ indicator fura-2 AM.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spatiotemporal Effects of Sonoporation Measured by Real-Time Calcium Imaging
R.E. Kumon
Ultrasound in Medicine & Biology, 35(3), 494-506 (2009)
Reciprocal Regulation of AMP-activated Protein Kinase and Phospholipase D*
Suman M
The Journal of Biological Chemistry, 290, 6986-6993 (2015)
Hakki Çelebi et al.
The International journal of oral & maxillofacial implants, 32(6), 1231–1240-1231–1240 (2017-05-19)
To compare the adhesion and proliferation effect of HGF-1 cells on pure titanium disks when the surfaces are unprocessed (machined surfaces after slicing) or modified with a laser. Twenty-eight titanium disks were divided into four groups. Three surface topographies were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service