6-Carboxytetramethylrhodamine (6-TAMRA) is used to create fluorescent DNA probes and labeled proteins/peptides. The corresponding succinimidyl ester is a single isomer fluorophore most commonly used in automated DNA sequencing.
The corresponding succinimidyl ester is a single isomer fluorophore most commonly used in automated DNA sequencing.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
This study introduces a modeling method for a supermolecular structure of microtubules for the development of a force generation material using motor proteins. 3D imaging by confocal laser scanning microscopy (CLSM) was used to obtain 3D volume density data. The
A novel fluorescent substrate was devised for the sirtuin (SIRT) class of human protein deacetylases comprised of a peptide sequence containing a single acetyl-lysine residue, with a fluorescent group (tetramethylrhodamine-6-carboxylic acid, 6-TAMRA) near the carboxyl terminus and a nonfluorescent quenching
Colloids and surfaces. B, Biointerfaces, 156, 414-421 (2017-05-30)
Light is an attractive trigger for release of active molecules from nanocarriers in biological systems. Here, we describe a phenomenon of light-induced release of a fluorescent dye from lipid nano-droplets under visible light conditions. Using auto-emulsification process we prepared nanoemulsion
Frontiers in veterinary science, 7, 625921-625921 (2021-02-27)
In the dog, implantation takes place at approximately 17 days of embryonal life and, while exposed to relatively high circulating progesterone concentrations, embryos presence is required for the formation of decidua. Furthermore, a balance between pro- and anti-inflammatory responses in
Organic-coated superparamagnetic iron oxide nanoparticles (OC-SPIONs) were synthesized and characterized by transmission electron microscopy and X-ray photoelectron spectroscopy. OC-SPIONs were transferred from organic media into water using poly(amidoamine) dendrimers modified with 6-TAMRA fluorescent dye and folic acid molecules. The saturation