Skip to Content
Merck

Skip To

5145

CytoSoft®

Elastic Moduli 64 kPa

Synonym(s):

CytoSoft Product, CytoSoft Technology

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
5 ea
Check Cart for Availability
$306.00

About This Item

UNSPSC Code:
12352202
EC Number:
231-130-8
NACRES:
NA.75
Form:
solid
Technique(s):
cell culture | mammalian: suitable

$306.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

irradiated

form

solid

packaging

pack of 5 plates

technique(s)

cell culture | mammalian: suitable

capacity

2.0-3.0 mL

size

6 wells

surface area

9.5 cm2

solubility

water: insoluble

General description

The rigidity of the substrate to which cells adhere can have a profound effect on cell morphology and gene expression. CytoSoft® products provide a tool to culture cells on substrates with various rigidities covering a broad physiological range. On the bottom of each well, there is a thin layer of specially formulated biocompatible silicone, whose elastic modulus (rigidity) is carefully measured. The surfaces of the gels in CytoSoft® products are functionalized to form covalent bonds with amines on proteins. This chemical functionalization is stable and the reaction does not require a catalyst, facilitating the coating of the gel surfaces with matrix proteins and plating cells.

The silicone substrates of CytoSoft® products are optically clear and have a low auto-florescence. The layer of silicone in each well is firmly bonded to the bottom of the well. Unlike hydrogels (such as polyacrylamide gels), silicone gels are not susceptible to hydrolysis, do not dry nor swell, are resilient and resistant to tearing or cracking, and their elastic moduli (rigidities) remain nearly unchanged during extended storage periods.

CytoSoft® products accommodate the harvesting of cells using enzymes such as trypsin and collagenase. There is no biochemical breakdown of the substrate during or after enzyme treatment, and there are no residuals of the substrate in the sample retrieved from a CytoSoft® plate.

This CytoSoft® product, Catalog Number 5145, has an elastic modulus of approximately 64 kPa in a standard flat-bottom 6-well plate. The thickness of the silicone gel is uniform with a ~0.5 mm thick layer of silicone in each well that is fully compatible with mammalian cell cultures. The plates are packaged individually, sterilized using irradiation and provided with 5 plates per package.

Legal Information

CytoSoft is a registered trademark of Advanced BioMatrix, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
514351425141
sterility

irradiated

sterility

irradiated

sterility

irradiated

sterility

irradiated

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

form

solid

form

solid

form

solid

form

solid

packaging

pack of 5 plates

packaging

pack of 5 plates

packaging

pack of 5 plates

packaging

pack of 5 plates

solubility

water: insoluble

solubility

water: insoluble

solubility

water: insoluble

solubility

water: insoluble

size

6 wells

size

6 wells

size

6 wells

size

6 wells


Still not finding the right product?

Explore all of our products under CytoSoft®


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

CytoSoft® modulus plates analyze substrate stiffness effects on cell behavior across a physiological range for mechanobiology research.





Global Trade Item Number

SKUGTIN
377031-250ML04061837751240
801553025004022536368078
5145-5EA04061832492957

Questions

Reviews

No rating value

Active Filters