Skip to Content
Merck
All Photos(1)

Key Documents

5141

Sigma-Aldrich

CytoSoft®

Elastic Moduli 2 kPa

Synonym(s):

CytoSoft Product, CytoSoft Technology

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$316.00

$316.00


Estimated to ship onMay 16, 2025


Request a Bulk Order

Select a Size

Change View
5 EA
$316.00

About This Item

EC Number:
UNSPSC Code:
12352202
NACRES:
NA.75

$316.00


Estimated to ship onMay 16, 2025


Request a Bulk Order

sterility

irradiated

form

solid

packaging

pack of 5 plates

technique(s)

cell culture | mammalian: suitable

capacity

2.0-3.0 mL

size

6 wells

surface area

9.5 cm2

solubility

water: insoluble

General description

The rigidity of the substrate to which cells adhere can have a profound effect on cell morphology and gene expression. CytoSoft® products provide a tool to culture cells on substrates with various rigidities covering a broad physiological range. On the bottom of each well, there is a thin layer of specially formulated biocompatible silicone, whose elastic modulus (rigidity) is carefully measured. The surfaces of the gels in CytoSoft® products are functionalized to form covalent bonds with amines on proteins. This chemical functionalization is stable and the reaction does not require a catalyst, facilitating the coating of the gel surfaces with matrix proteins and plating cells.

The silicone substrates of CytoSoft® products are optically clear and have a low auto-florescence. The layer of silicone in each well is firmly bonded to the bottom of the well. Unlike hydrogels (such as polyacrylamide gels), silicone gels are not susceptible to hydrolysis, do not dry nor swell, are resilient and resistant to tearing or cracking, and their elastic moduli (rigidities) remain nearly unchanged during extended storage periods.

CytoSoft® products accommodate the harvesting of cells using enzymes such as trypsin and collagenase. There is no biochemical breakdown of the substrate during or after enzyme treatment, and there are no residuals of the substrate in the sample retrieved from a CytoSoft® plate.

This CytoSoft® product, Catalog Number 5141, has an elastic modulus of approximately 2 kPa in a standard flat-bottom 6-well plate. The thickness of the silicone gel is uniform with a ~0.5 mm thick layer of silicone in each well that is fully compatible with mammalian cell cultures. The plates are packaged individually, sterilized using irradiation and provided with 5 plates per package.

Legal Information

CytoSoft is a registered trademark of Advanced BioMatrix, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service