Skip to Content
Merck
All Photos(1)

Key Documents

45299

Sigma-Aldrich

Epoxide Hydrolase from Rhodococcus rhodochrous

lyophilized powder, beige, ≥0.2 U/g

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

lyophilized powder

specific activity

≥0.2 U/g

color

beige

storage temp.

2-8°C

Biochem/physiol Actions

Epoxide hydrolase is an enantioselective catalyst for the hydrolytic kinetic resolution of epoxides.[1]

Unit Definition

1 U corresponds to the amount of enzymes which hydrolizes 1 μmol (S)-NEPC [(2S,3S)-trans-3-phenyl-2-oxiranylmethyl-4-nitrophenyl carbonate, Prod. No. 04088] per minute at pH 8.0 and 25 °C

Other Notes

Asymmetric hydrolysis of epoxides to optically active diols[2][3]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mischitz, M., et al.
Tetrahedron Asymmetry, 7, 2041-2041 (1996)
Hechtberger, P.
Tetrahedron Asymmetry, 4, 1161-1161 (1993)
E N Jacobsen
Accounts of chemical research, 33(6), 421-431 (2000-07-13)
The discovery of the metal salen-catalyzed asymmetric ring-opening (ARO) of epoxides is chronicled. A screening approach was adopted for the identification of catalysts for the addition of TMSN(3) to meso-epoxides, and the chiral (salen)CrN(3) complex was identified as optimal. Kinetic
Ana W Y Cheong et al.
Biology of reproduction, 81(1), 126-132 (2009-03-27)
Somatic cell-embryo coculture enhances embryo development in vitro by producing embryotrophic factor(s) and/or removing harmful substances from the culture environment. Yet, the underlying molecular mechanisms on how somatic cells remove the toxicants from the culture medium remain largely unknown. By
Zhuo Shao et al.
Arteriosclerosis, thrombosis, and vascular biology, 34(3), 581-586 (2014-01-25)
Regulation of angiogenesis is critical for many diseases. Specifically, pathological retinal neovascularization, a major cause of blindness, is suppressed with dietary ω3-long-chain polyunsaturated fatty acids (ω3LCPUFAs) through antiangiogenic metabolites of cyclooxygenase and lipoxygenase. Cytochrome P450 epoxygenases (CYP2C8) also metabolize LCPUFAs

Questions

  1. Hello, I was wondering if you could please let me know the molecular weight of this enzyme from Rhodococcus rhodochrous.

    1 answer
    1. The molecular weight of this product has not been determined. Various publications suggest that this enzyme is a dimer with an approximate molecular weight of 110,000 daltons. Each subunit is reported to be 57,000 daltons. See the link below for additional information:
      https://pubmed.ncbi.nlm.nih.gov/7117244/

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service