Skip to Content
Merck
All Photos(1)

Key Documents

44511

Sigma-Aldrich

Sodium D-galactonate

≥98.0% (TLC)

Synonym(s):

D-Galactonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O7 · xNa+
Molecular Weight:
196.16 (free acid basis)
Beilstein:
1726059
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

≥98.0% (TLC)

form

powder

storage temp.

2-8°C

SMILES string

OC[C@@H](O)[C@H](O)[C@H](O)[C@@H](O)C(O)=O

InChI

1S/C6H12O7/c7-1-2(8)3(9)4(10)5(11)6(12)13/h2-5,7-11H,1H2,(H,12,13)/t2-,3+,4+,5-/m1/s1

InChI key

RGHNJXZEOKUKBD-MGCNEYSASA-N

Biochem/physiol Actions

Metabolite in galactose metabolim, non-phosphorylative Entner-Doudoroff pathway, D-galactonate degradation, De Ley-Doudoroff pathway, semi-phosphorylative Entner-Doudoroff pathway

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Schadewaldt et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 801(2), 249-255 (2004-01-31)
A stable isotope dilution assay was developed for the sensitive determination of D-galactonic acid. D-[U-13C(6)]galactono-1,4-lactone was prepared as internal standard. Unlabelled and U-13C-labelled D-galactonic acid species were converted to the N-(1-butyl)galactonamide pentaacetate derivatives and assessed by gas chromatography-mass spectrometry (GC-MS).
T Shinka et al.
Journal of chromatography. B, Biomedical sciences and applications, 732(2), 469-477 (1999-10-12)
We determined urinary galactose and 4-hydroxyphenyllactic acid (4HPLA) in 4338 of 5-day-old newborns using a newly developed GC-MS screening method. Fifty-two infants were chemically diagnosed as having transient galactosuria based upon elevated urinary galactose levels (4.78-30.53 mg/mg creatinine, control 1.10
Claire T Yager et al.
Molecular genetics and metabolism, 80(3), 283-289 (2003-12-19)
The red blood cell (RBC) concentration of galactitol and galactonate was measured in 27 patients with galactose-1-phosphate uridyltransferase (GALT) deficiency galactosemia and 19 non-galactosemic subjects by a newly devised isotope dilution gas chromatography/mass spectrometry (GC/MS) method. The method utilizing UL[13C]galactitol
J D Shoemaker et al.
Journal of chromatography, 562(1-2), 125-138 (1991-01-02)
Eighty-five clinical urine samples and nineteen urine samples previously found by other laboratories to suggest genetic metabolic defects were prepared for trimethylsilylation by treatment with urease, followed by azeotropic dehydration. The "Target Analyte Search" program provided with the VG Trio

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service