Merck
  • Automated screening of urine samples for carbohydrates, organic and amino acids after treatment with urease.
Automated screening of urine samples for carbohydrates, organic and amino acids after treatment with urease.
Journal of chromatography (1991-01-02)
J D Shoemaker, W H Elliott
ABSTRACT
Eighty-five clinical urine samples and nineteen urine samples previously found by other laboratories to suggest genetic metabolic defects were prepared for trimethylsilylation by treatment with urease, followed by azeotropic dehydration. The "Target Analyte Search" program provided with the VG Trio 2 gas chromatograph-mass spectrometer required 6 min to quantify 103 compounds relative to endogenous urinary creatinine. This technique has been used to confirm diagnoses including cystinuria, lysinuria, medium-chain acyldehydrogenase deficiency, ornithine transcarbamylase deficiency, aspartylglucosaminuria, methylmalonic, propionic and glutaric acidurias.
Product Description
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
L-Asparagine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Sigma-Aldrich
L-Glutamine
Pricing and availability is not currently available.
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylpyruvic acid, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Fumaric acid, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Methylmalonic acid, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylmalonic acid, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Pricing and availability is not currently available.
Sigma-Aldrich
Adipic acid, 99%
Pricing and availability is not currently available.
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Maleic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Sigma-Aldrich
Suberic acid, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Pricing and availability is not currently available.
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Pricing and availability is not currently available.
Supelco
Fumaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Pricing and availability is not currently available.