Skip to Content
Merck
All Photos(1)

Documents

42030

Sigma-Aldrich

3,5-Dinitrobenzoyl chloride

for fluorescence, ≥98.0% (AT)

Synonym(s):

DNBC, NSC 2697

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3COCl
CAS Number:
Molecular Weight:
230.56
Beilstein:
990249
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

grade

for fluorescence

vapor density

7.6 (vs air)

Assay

≥98.0% (AT)

form

solid

bp

196 °C/11 mmHg (lit.)

mp

68-69 °C (lit.)
68-71 °C

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1cc(cc(c1)[N+]([O-])=O)C(Cl)=O

InChI

1S/C7H3ClN2O5/c8-7(11)4-1-5(9(12)13)3-6(2-4)10(14)15/h1-3H

InChI key

NNOHXABAQAGKRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the UV-visible absorption derivatization of alcohols in liquid chromatography

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Markowska et al.
Roczniki Akademii Medycznej w Bialymstoku (1995), 42(1), 129-140 (1997-01-01)
A carbocyclic analogue of distamycin was obtained, in which the N-methylpyrrole rings were substituted by disubstituted benzene rings. Additionally, N-chloro- or N-bromoacetyl groups, displaying alkylating properties, were introduced. The synthesis, starting from 3,5-dinitrobenzoyl chloride, consisted of five stages.
S Wongyai et al.
Biomedical chromatography : BMC, 2(6), 254-257 (1988-11-01)
Analytical methods are described for the quantitative determination of putrescine, cadaverine, spermidine, spermine and the acetylated derivatives of spermidine and spermine in biological fluids using pre-column derivatization with either benzoyl chloride or 3,5-dinitrobenzoyl chloride, which were added to each sample
Jiu-gao Yu et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 38(3), 191-195 (2003-07-02)
To prepare a novel creatinine adsorbent with anti-renal failure activity. A novel starch ester was prepared under heterogeneous condition by the reaction of starch (ST) and 3, 5-dinitrobenzoyl chloride (DNBZ-Cl). The products were characterized by means of elemental analysis, FTIR
T. Jupille
Journal of Chromatographic Science, 17, 160-160 (1979)
J Kirschbaum et al.
Journal of chromatography. A, 881(1-2), 517-530 (2000-07-25)
The reagent 3,5-dinitrobenzoyl chloride (DNBZ-Cl) was tested for pre-column derivatization of biogenic amines (BAs). Samples were derivatized within 3 min in 1 M NaOH at ambient temperature by adding 2-propanole and 50 mM DNBZ-Cl in acetonitrile. The reaction was terminated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service