Skip to Content
Merck
All Photos(1)

Key Documents

M2876

Sigma-Aldrich

(±)-α-Methoxyphenylacetic acid

crystalline

Synonym(s):

O-Methyl-DL-mandelic acid, MOPA

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
$85.66
5 G
$101.00
10 G
$195.00
25 G
$433.00

$85.66

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
2 G
$85.66
5 G
$101.00
10 G
$195.00
25 G
$433.00

About This Item

Linear Formula:
C6H5CH(OCH3)COOH
CAS Number:
Molecular Weight:
166.17
Beilstein:
2209197
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$85.66

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

≥99.0%

form

crystalline

bp

165 °C ((lit.))
165 °C/18 mmHg (lit.)

mp

69-71 °C (lit.)
69-71 °C

SMILES string

COC(C(O)=O)c1ccccc1

InChI

[1S/C9H10O3/c1-12-8(9(10)11)7-5-3-2-4-6-7/h2-6,8H,1H3,(H,10,11)]
1S/C9H10O3/c1-12-8(9(10)11)7-5-3-2-4-6-7/h2-6,8H,1H3,(H,10,11)

InChI key

[DIWVBIXQCNRCFE-UHFFFAOYSA-N]
DIWVBIXQCNRCFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(±)-α-Methoxyphenylacetic acid, also known as MOPA is commonly used as a resolving agent and building block in various organic synthesis.

Application

(±)-α-Methoxyphenylacetic acid can be used as a reactant to synthesize:      
  • N






  • -(Cyclohexylmethyl)-2-methoxy-2-phenylethylamine by reacting with cyclohexylmethylamine in the presence of EDCI/HOBt/Et3N.      
  • 4-(methoxyphenylmethyl)-2-methylpyridine by reacting with 2-methyl-4-pyridinecarbonitrile via decarboxylative arylation reaction in the presence of a photocatalyst.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shoko Yamazaki et al.
The Journal of organic chemistry, 82(13), 6748-6763 (2017-06-13)
Catalytic cyclization of amides of ethenetricarboxylate bearing ether and acetal groups has been examined. The reaction of the amides bearing cyclic ether and acetal groups in the presence of Lewis acid such as Sc(OTf)3 gave spirocyclic piperidine derivatives as major
Yiyang Chen et al.
Organic letters, 21(7), 2130-2133 (2019-03-13)
1-(4-(9 H-Carbazol-9-yl)phenyl)-3-amino-9 H-fluorene-2,4-dicarbonitrile as a new photocatalyst for the decarboxylative cross-coupling reaction of α-amino acids or α-oxy carboxylic acids with arylnitriles is described. This light-driven reaction enables a variety of benzylic amines and ethers to be prepared from readily available
Bin Chang et al.
Journal of clinical microbiology, 53(10), 3318-3324 (2015-08-14)
Streptococcus pneumoniae colonizes the nasopharyngeal mucus in healthy people and causes otitis media, pneumonia, bacteremia, and meningitis. In this study, we analyzed an S. pneumoniae strain that caused 7 repeated pneumonia episodes in an 80-month-old patient with cerebral palsy during
Radoslaw Laufer et al.
Bioorganic & medicinal chemistry, 22(17), 4968-4997 (2014-07-22)
TTK kinase was identified by in-house siRNA screen and pursued as a tractable, novel target for cancer treatment. A screening campaign and systematic optimization, supported by computer modeling led to an indazole core with key sulfamoylphenyl and acetamido moieties at
Weidong Liu et al.
Chembiochem : a European journal of chemical biology, 16(6), 924-929 (2015-03-11)
A meso-diaminopimelate dehydrogenase (DAPDH) from Clostridium tetani E88 (CtDAPDH) was found to have low activity toward the D-amino acids other than its native substrate. Site-directed mutagenesis similar to that carried out on the residues mutated by Vedha-Peters et al. resulted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service