Skip to Content
Merck
All Photos(1)

Key Documents

30960

Sigma-Aldrich

Deoxycholic acid

≥99.0% (T)

Synonym(s):

3α,12α-Dihydroxy-5β-cholanic acid, 7-Deoxycholic acid, Desoxycholic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
Beilstein:
3219882
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

biological source

ox bile

Quality Level

description

anionic

Assay

≥99.0% (T)

form

powder

optical activity

[α]20/D +54±1°, c = 1% in ethanol

mol wt

392.57 g/mol

impurities

≤1% cholic acid (TLC)

mp

170-175 °C
171-174 °C (lit.)

functional group

carboxylic acid

SMILES string

C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C24H40O4/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3/h14-21,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-/m1/s1

InChI key

KXGVEGMKQFWNSR-LLQZFEROSA-N

Gene Information

human ... ATIC(471)

Looking for similar products? Visit Product Comparison Guide

General description

Deoxycholic acid is an amphiphilic molecule and an important constituent of bile acid. It contains hydroxyl groups substituted at the 3α and 12α positions and a carboxyl group towards the end of the branched-chain. These moieties contribute to the hydrophilic nature of the molecule and the hydrophobic cyclopentanophenanthrene forms its center.[1]

Application

Deoxycholic acid has been used as:
  • a hyphal-inhibitory compound in an ex vivo assay to study the hyphal morphogenesis of Candida albicans in the gastrointestinal tract[2]
  • an internal standard to estimate the toxin concentration in the cell pellet[3]
  • a bile acid (BA) standard to quantitatively measure BA in various samples from treated mice by ultra-performance liquid chromatography triple quadrupole mass spectrometry[4]

Biochem/physiol Actions

Deoxycholic acid, due to its amphiphilicity, significantly helps to solubilize, emulsify, and absorb fat, vitamins, and cholesterol in the body.[1] High levels of intestinal deoxycholic acid might cause colorectal cancer by inducing oxidative stress and leading to DNA damage. It acts as an oncogene and pro-tumor factor.[5]

Other Notes

For the solubilization of the β-adrenergic receptor[6]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and characterizations of a novel deoxycholic acid-O-carboxymethylated chitosan-folic acid conjugates and self-aggregates
Wang F, et al.
Carbohydrate Polymers, 84, 1192-1200 (2011)
E Nedivi et al.
The Journal of biological chemistry, 259(9), 5803-5808 (1984-05-10)
Agonist, but not antagonist, protects the beta-adrenergic receptor from inactivation during solubilization in deoxycholate. Protection is apparently due to locking of the agonist in the receptor, a high affinity interaction induced or stabilized by this detergent. The guanyl nucleotide-binding protein
Thierry Claudel et al.
Arteriosclerosis, thrombosis, and vascular biology, 25(10), 2020-2030 (2005-07-23)
Bile acids are the end products of cholesterol metabolism. They are synthesized in the liver and secreted via bile into the intestine, where they aid in the absorption of fat-soluble vitamins and dietary fat. Subsequently, bile acids return to the
Jeffery L Smith et al.
Hepatology (Baltimore, Md.), 39(6), 1673-1682 (2004-06-09)
Focal biliary cirrhosis causes significant morbidity and mortality in cystic fibrosis (CF). Although the mechanisms of pathogenesis remain unclear, bile acids have been proposed as potential mediators of liver injury. This study examined bile acid composition in CF and assessed
J Stadler et al.
Cancer letters, 38(3), 315-320 (1988-01-01)
Rectal biopsies and fecal collections were obtained from a consecutive series of 34 outpatients prior to colonoscopy at a gastroenterology clinic. Subsequently, 14 were found to have no colonic pathology, 13 had adenomatous polyps, (3 of those had a previous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service