![Deoxycholic acid certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/133/075/bb38155a-4c3d-4225-97de-14fbf5cbf5ee/640/bb38155a-4c3d-4225-97de-14fbf5cbf5ee.png)
Deoxycholic acid
![Deoxycholic acid certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/133/075/bb38155a-4c3d-4225-97de-14fbf5cbf5ee/640/bb38155a-4c3d-4225-97de-14fbf5cbf5ee.png)
Synonym(s):
3α,12α-Dihydroxy-5β-cholanic acid, 7-Deoxycholic acid, Desoxycholic acid
Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Tissue
Quality Segment
Functional Group
Application
Package Size
Special Grade
Showing 1-4 of 4