Skip to Content
Merck
All Photos(1)

Documents

243M-1

Sigma-Aldrich

Desmin (D33) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

D33, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (243M-14)
vial of 0.5 mL concentrate (243M-15)
bottle of 1.0 mL predilute (243M-17)
vial of 1.0 mL concentrate (243M-16)
bottle of 7.0 mL predilute (243M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG1κ

control

muscle

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... DES(1674)

General description

Anti-desmin detects a protein that is expressed by cells of normal smooth, skeletal, and cardiac muscles. The light microscope has suggested that desmin is primarily located at or near the periphery of Z lines in striated muscle fibrils. In smooth muscle, desmin interconnects cytoplasmic dense bodies with membrane bound dense plaques. Anti-desmin reacts with leiomyomas, leiomyosarcoma, rhabdomyomas, rhabdomyosarcoma, and perivascular cells of glomus tumors of the skin. This antibody is used to demonstrate the myogenic components/derivation of tumors. Desmin can also be present in myofibroblasts and be focally positive in desmoid fibromatosis.

Quality


IVD

IVD

IVD

RUO

Linkage

Desmin Positive Control Slides, Product No. 243S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Altmannsberger et al.
The American journal of pathology, 118(1), 85-95 (1985-01-01)
Putative human rhabdomyosarcoma (RMS) has been divided into two groups according to desmin content. Twenty-five tumors with histologic features consistent with but not necessarily sufficient to prove a diagnosis of RMS were desmin-positive. More than 95% of the tumor cells
Umio Yamaguchi et al.
Virchows Archiv : an international journal of pathology, 445(2), 142-150 (2004-07-03)
To confirm the usefulness of an immunohistochemical panel of antibodies for KIT (c-kit/CD117), CD34, desmin, smooth-muscle actin (SMA), h-caldesmon (HCD), S-100 protein, neuron-specific enolase (NSE), and beta-catenin, 297 mesenchymal and peripheral nerve-sheath tumors of the gastrointestinal tract and intra-abdominal locations
E Debus et al.
The EMBO journal, 2(12), 2305-2312 (1983-01-01)
A set of monoclonal antibodies to desmin has been isolated from a fusion of mouse myeloma cells with spleen cells from mice immunized with purified porcine desmin. Eleven group I antibodies recognized desmin in the immune blot, and using defined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service