Skip to Content
Merck
All Photos(1)

Key Documents

195855

Sigma-Aldrich

Precocene I

99%

Synonym(s):

7-Methoxy-2,2-dimethyl-3-chromene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O2
CAS Number:
Molecular Weight:
190.24
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.25

Assay

99%

form

liquid

refractive index

n20/D 1.56 (lit.)

bp

68 °C/0.1 mmHg (lit.)

density

1.052 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

SMILES string

COc1ccc2C=CC(C)(C)Oc2c1

InChI

1S/C12H14O2/c1-12(2)7-6-9-4-5-10(13-3)8-11(9)14-12/h4-8H,1-3H3

InChI key

CPTJXGLQLVPIGP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Venugopal et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 127(2), 153-163 (2000-11-18)
Investigations were carried out to determine the role of juvenile hormone (JH) and 20-hydroxy ecdysone in the synthesis and uptake of vitellogenins, which were earlier identified, purified and characterised, in Dysdercus koenigii. The concentration(s) of vitellogenin(s) in fat body, haemolymph
P De Azambuja et al.
Memorias do Instituto Oswaldo Cruz, 86 Suppl 2, 113-115 (1991-01-01)
Proallatotoxins, and particularly precocenes, are exceptionally promising models for studying Rhodnius prolixus physiology and for comparison with other natural compounds with antihormonal activities. Effects of precocenes on feeding, development and reproduction of R. prolixus are being detailed. The precocenes reveal
Maryla Szczepanik et al.
Journal of agricultural and food chemistry, 53(15), 5905-5910 (2005-07-21)
Starting from precocenes I and II, four of their derivatives with a lactone moiety were obtained. The compounds have been assessed as antifeedants against several diverse insect species including the storage pests the confused flour beetle (Tribolium confusum Duv., larvae
Carlos A Gartner et al.
Biochemistry, 44(6), 1846-1855 (2005-02-09)
Cytochrome P450 3A4 is a drug-metabolizing enzyme of extraordinarily broad substrate specificity. This quality imparts upon the enzyme special importance in understanding its determinants of activity and substrate recognition. Limited successes in P450 3A4 active-site structure studies have been achieved
S J Chiou et al.
Insect biochemistry and molecular biology, 28(10), 801-808 (1998-11-10)
The cDNA coding for a Ser-protease-related protein (Scg-SPRP) was cloned from desert locust (Schistocerca gregaria) midgut. The derived amino acid sequence consists of 260 residues and shows strong sequence similarity to insect trypsin-like molecules. It is, however, likely that Scg-SPRP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service