Skip to Content
Merck
All Photos(1)

Key Documents

206512

Sigma-Aldrich

1-Bromo-4-ethynylbenzene

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$127.00

$127.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$127.00

About This Item

Linear Formula:
HC≡CC6H4Br
CAS Number:
Molecular Weight:
181.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$127.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

mp

64-67 °C (lit.)

functional group

bromo

SMILES string

Brc1ccc(cc1)C#C

InChI

1S/C8H5Br/c1-2-7-3-5-8(9)6-4-7/h1,3-6H

InChI key

LTLVZQZDXQWLHU-UHFFFAOYSA-N

Related Categories

Application

1-Bromo-4-ethynylbenzene was used in the synthesis of 1-bromo-4-(4′-tetrahydropyranyloxy-1′-ethynylphenyl)benzene[1].

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long TM and Swager TM.
Journal of Materials Chemistry, 12(12), 3407-3412 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service