Skip to Content
Merck
All Photos(1)

Key Documents

14530C

SAFC

EX-CELL® EBx GRO-I Serum-Free Media

Animal-component free, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

EX-CELL® EBx Serum-Free Media

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$75.00
14530C-1000ML
$130.00

$75.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
500 ML
$75.00
14530C-1000ML
$130.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

$75.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


description

Animal-component-free, low protein
for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

availability

not available in Japan

technique(s)

cell culture | mammalian: suitable
cell culture | stem cell: suitable

components

L-glutamine: no

shipped in

ambient

storage temp.

2-8°C
2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

EX-CELL® EBx GRO-I Serum-Free Media has been used to culture EB66 cell line derived from duck embryonic stem cells.[1]

Legal Information

EBx is a trademark of Vivalis SA
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

EB66 cell line, a duck embryonic stem cell-derived substrate for the industrial production of therapeutic monoclonal antibodies with enhanced ADCC activity
Stephane Olivier
MAbs, 2(4) (2010)

Related Content

Nancy-520 for DNA Detection and Quantitation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service