Skip to Content
Merck
All Photos(1)

Key Documents

12685

Sigma-Aldrich

Nω-Phospho-L-arginine lithium salt hydrate

≥95.0% (TLC)

Synonym(s):

H-Arg(PO3H2)-OH lithium salt, N5-(Phosphonoamidino)-L-ornithine lithium salt, N5-[Imino(phosphonoamino)methyl]-L-ornithine lithium salt, L-2-Amino-5-(N′-phosphonoguanidino)valeric acid lithium salt, Lithium L-arginine phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15N4O5P · xLi+ · yH2O
CAS Number:
Molecular Weight:
254.18 (free acid basis)
EC Number:
UNSPSC Code:
12352209
NACRES:
NA.26

Assay

≥95.0% (TLC)

form

powder or crystals

color

white to off-white

storage temp.

−20°C

InChI

1S/C6H15N4O5P/c7-4(5(11)12)2-1-3-9-6(8)10-16(13,14)15/h4H,1-3,7H2,(H,11,12)(H5,8,9,10,13,14,15)/t4-/m0/s1

InChI key

CCTIOCVIZPCTGO-BYPYZUCNSA-N

Biochem/physiol Actions

Important metabolite in arginine and proline metabolism, high-energy metabolite, constituent of crustaceans and crayfish muscle.

Analysis Note

may contain 8% more water then theoretically in monohydrate expected

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaspar E Canepa et al.
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology, 160(1), 40-43 (2011-06-01)
Phytomonas are trypanosomatid plant parasites closely related to parasites that cause several human diseases. Little is known about the biology of these organisms including aspects of their metabolism. Arginine kinase (E.C. 2.7.3.3) is a phosphotransferase which catalyzes the interconversion between
Omar Davulcu et al.
Biochemistry, 50(19), 4011-4018 (2011-03-24)
Arginine kinase catalyzes the reversible transfer of a phosphoryl group between ATP and arginine. It is the arthropod homologue of creatine kinase, buffering cellular ATP levels. Crystal structures of arginine kinase, in substrate-free and substrate-bound forms, have revealed large conformational
Karina D García-Orozco et al.
International archives of allergy and immunology, 144(1), 23-28 (2007-05-15)
Consumption of seafood can produce allergic symptoms in susceptible individuals and crustacean allergies are the most frequently reported causes of allergic reactions. An allergen from the muscle of the white shrimp Litopenaeus vannamei was purified by ion exchange chromatography and
Margaret Werr et al.
Insect biochemistry and molecular biology, 39(9), 634-645 (2009-07-15)
Arginine kinase (ATP:l-arginine omega-N-phosphotransferase, EC2.7.3.3.; AK) is an enzyme crucial for the energy metabolism of insects and other invertebrates, that has known allergenic potential in humans and that has been proposed as a pesticidal drug target. Here we report the
Jonathan Bragg et al.
Journal of bacteriology, 194(10), 2668-2676 (2012-03-06)
Arginine kinases catalyze the reversible transfer of a high-energy phosphoryl group from ATP to l-arginine to form phosphoarginine, which is used as an energy buffer in insects, crustaceans, and some unicellular organisms. It plays an analogous role to that of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service