Skip to Content
Merck
All Photos(3)

Key Documents

I5513

Sigma-Aldrich

Imidazole

Imidazole
1 of 1 reviewers received a sample product or took part in a promotion

for molecular biology, ≥99% (titration)

Synonym(s):

3-Azapyrrole, 1,3-Diaza-2,4-cyclopentadiene, Glyoxaline

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$208.00
25 G
$671.00
100 G
$1,980.00

$208.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
5 G
$208.00
25 G
$671.00
100 G
$1,980.00

About This Item

Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
Beilstein:
103853
EC Number:
MDL number:
UNSPSC Code:
12352005
eCl@ss:
39161001
PubChem Substance ID:
NACRES:
NA.25

$208.00


In StockDetails


Request a Bulk Order

grade

for molecular biology

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99% (titration)

form

crystalline powder

storage condition

dry at room temperature

technique(s)

electrophoretic mobility shift assay: suitable
molecular cloning: suitable
protein purification: suitable

color

white

pH

6.2-7.8 (25 °C)

pKa (25 °C)

6.95

bp

256 °C (lit.)

mp

88-91 °C (lit.)

solubility

water: 500 mg/mL, clear, colorless to very faintly yellow

density

1.004 g/cm3 at 20—25 °C
1.006 g/cm3 at 20—25 °C

suitability

suitable for molecular biology

application(s)

diagnostic assay manufacturing
genomic analysis
life science and biopharma
microbiology

foreign activity

DNase, RNase and protease, none detected

storage temp.

room temp

SMILES string

c1c[nH]cn1

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)

InChI key

RAXXELZNTBOGNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Imidazole, also known as Glyoxalin, is an aromatic heterocyclic diazole and a key biological buffer with a pH range of 6.2 to 7.8 at 25°C. It has a wide range of applications in molecular biology, biochemistry, and proteomics. In proteomics, imidazole is essential for eluting histidine-tagged proteins from IMAC columns. It is a strong chelator of divalent cations, such as nickel and zinc, which disrupts the interaction between the His-tag and the metal ions. This allows for the efficient elution of purified proteins.

Furthermore, imidazole is indispensable for enhancing protein purity. It is widely used in preparing buffer solutions optimized for downstream purification columns. Imidazole solutions are also effective elution and washing agents throughout protein purification processes. Additionally, imidazole can be used for assays of horseradish peroxidase (HRP) and reverse staining of SDS-PAGE gels for protein detection. Imidazole is a versatile and valuable reagent for a wide range of biological applications. It is an essential component of many buffers and is also used in protein purification and detection protocols.

Application

Imidazole has been used

  • in the lysis, wash and elution buffer for the purification of histidine tagged Sonic Hedgehog(shh-N) protein[1]
  • in elution buffer in stepwise gradient for the purification of histidine tagged aldo keto reductases using nickel affinity chromatography[2]
  • as a component of homogenization buffer for the purification of phagosomal compartments from dendritic cells[3]
  • as a binding buffer, wash buffer, and elution buffer in a study involving protein expression and purification of bioactive growth factors for use in cell culture and cellular agriculture[4][3]
Excellent for buffers in the range of pH 6.2-7.8

Biochem/physiol Actions

Imidazole derivatives have antibacterial, antifungal and anticancer functionality.[5] It interacts with DNA and also binds to protein and stops cell division. It also acts as a microtubule destabilizing agents and inhibits topoisomerase and Cytochrome P450 Family 26 Subfamily A Member 1 (CYP26A1) enzymes. Imidazole based anticancer drug find applications in cancer chemotherapy.[6] It is used as buffer component for purification of the histidine tagged recombinant proteins in immobilized metal-affinity chromatography (IMAC).[7]

Features and Benefits

  • Suitable for Molecular Biology and Biochemical Research
  • Can be used as a Buffer component, for Protein separation
  • Free from DNase, NICKase, RNase, and Protease
  • Effective Buffering from pH 6.2 to 7.8 (25 °C) with a pKa of 6.9 (25 °C)

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein expression and purification of bioactive growth factors for use in cell culture and cellular agriculture
Semper C et al.
STAR protocols, 4, 102351-102351 (2023)
Molecular pathways regulating pro-migratory effects of Hedgehog signaling
Hochman E, et al.
The Journal of Biological Chemistry, 281(45), 33860-33870 (2006)
Imidazole: Having versatile biological activities
Verma A, et al.
Journal of Chemistry, 2013 (2013)
Functional expression of novel human and murine AKR1B genes
Salabei JK, et al.
Chemico-Biological Interactions, 191(1-3), 177-184 (2011)
Imidazoles as potential anticancer agents
Ali I, et al.
MedChemComm, 8(9), 1742-1773 (2017)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Atlanta, GA
    • Review 1
    • Vote 1
    5 out of 5 stars.

    SARS-CoV-2 Spike and RBD protein purification

    The product quality is very good and helped to purify SARS-CoV-2 RBD and Spike protein

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service