Skip to Content
Merck
All Photos(1)

Key Documents

123-100

Sigma-Aldrich

Attachment Factor Solution (100 ML)

Synonym(s):

AFS

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$43.10

$43.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$43.10

About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

$43.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

bovine

form

liquid

shelf life

1 yr

packaging

pkg of 100 mL

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Attachment factor solution (AFS) is a sterile-filtered solution of attachment factor. It is used as a substrate for plating and spreading endothelial cells. AFS is used for coating cell culture flasks, dishes, and multi-well plates.

Application

Attachment factor solution (100 ML) has been used to coat cultural flasks prior to brain microvascular endothelial cell culture.[1]

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han Ly et al.
Annals of neurology, 82(2), 208-222 (2017-07-12)
The brain blood vessels of patients with type 2 diabetes and dementia have deposition of amylin, an amyloidogenic hormone cosecreted with insulin. It is not known whether vascular amylin deposition is a consequence or a trigger of vascular injury. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service