Skip to Content
Merck
All Photos(1)

Key Documents

08717

Supelco

Atto 594 maleimide

BioReagent, suitable for fluorescence, ≥90% (Coupling efficiency with N-Acetylcysteine, HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$427.00

$427.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$427.00

About This Item

MDL number:
UNSPSC Code:
12352111
NACRES:
NA.32

$427.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

product line

BioReagent

Assay

≥90% (Coupling efficiency with N-Acetylcysteine, HPLC)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

transmittance

254 nm
604 nm

fluorescence

λex 601 nm; λem 627 nm in 0.1 M phosphate pH 7.0

λ

in ethanol (with 0.1% trifluoroacetic acid)

suitability

suitable for fluorescence

detection method

fluorometric

storage temp.

−20°C

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Semi-Solid o/w Emulsions Based on Sucrose Stearates: Influence of Oil and Surfactant Type on Morphology and Rheological Properties.
Klang, V., et al.
Journal of Dispersion Science and Technology, 34, 322-333 (2013)
Massa J Shoura et al.
Nucleic acids research, 40(15), 7452-7464 (2012-05-17)
The Cre-recombination system has become an important tool for genetic manipulation of higher organisms and a model for site-specific DNA-recombination mechanisms employed by the λ-Int superfamily of recombinases. We report a novel quantitative approach for characterizing the probability of DNA-loop
Michael R Williams et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(27), 9228-9237 (2012-07-06)
The potassium channel Kv1.2 α-subunit is expressed in cerebellar Purkinje cell (PC) dendrites where its pharmacological inhibition increases excitability (Khavandgar et al., 2005). Kv1.2 is also expressed in cerebellar basket cell (BC) axon terminals (Sheng et al., 1994), where its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service