Skip to Content
Merck

Skip To

08602

Nigerose

≥90.0% (HPLC)

Synonym(s):

3-O-α-D-Glucopyranosyl-D-glucose, Sakebiose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
$240.00

About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
MDL number:

$240.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90.0% (HPLC)

form

solid

color

white

storage temp.

room temp

InChI

1S/C12H22O11/c13-1-3-5(15)7(17)8(18)12(22-3)23-10-6(16)4(2-14)21-11(20)9(10)19/h3-20H,1-2H2/t3-,4-,5-,6-,7+,8-,9-,10+,11?,12-/m1/s1

InChI key

QIGJYVCQYDKYDW-NSYYTRPSSA-N

Application

Nigerose (Sakebiose), an α(1,3) linked glucose disaccaharide, is used as a substrate to identify, differentiate and quantify enzymes such as nigerose phosphorylase(s) and α-glucosidase(s).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
0538992145K4769
assay

≥90.0% (HPLC)

assay

≥98.0% (HPLC)

assay

≥97% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

200

form

solid

form

-

form

-

form

powder

storage temp.

room temp

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C

color

white

color

-

color

-

color

white to off-white


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters