Skip to Content
Merck
All Photos(1)

Key Documents

04685

Sigma-Aldrich

Nε-Methyl-L-lysine hydrochloride

≥98.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16N2O2 · HCl
CAS Number:
Molecular Weight:
196.68
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32
grade:
for analytical purposes

grade

for analytical purposes

Quality Level

Assay

≥98.0% (TLC)

optical activity

[α]/D 20.5±1.5°, c = 0.1 in 1 M HCl

storage temp.

2-8°C

SMILES string

Cl.CNCCCC[C@H](N)C(O)=O
Cl.CNCCCC[C@H](N)C(O)=O

InChI

1S/C7H16N2O2.ClH/c1-9-5-3-2-4-6(8)7(10)11;/h6,9H,2-5,8H2,1H3,(H,10,11);1H/t6-;/m0./s1

InChI key

AQELUQTVJOFFBN-RGMNGODLSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

N ε-methyl-L-lysine was identified as a lysine analog with inhibitory effects on the growth and sporulation of Penicillium chrysogenum and benzyl-penicillin formation by mycelia.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A genetically encoded epsilon-N-methyl lysine in mammalian cells.
Dan Groff et al.
Chembiochem : a European journal of chemical biology, 11(8), 1066-1068 (2010-04-28)
M Friedman et al.
The Journal of nutrition, 111(8), 1362-1369 (1981-08-01)
Growth assays using mice on synthetic amino acid diets showed that substituting epsilon-N-methyl-L-lysine, epsilon-N-dimethyl-L-lysine and epsilon-N-trimethyl-L-lysine for lysine resulted in relative replacement values about 1/12, 1/20 and 1/25, respectively, of that obtained with the standard lysine diet. Similar studies showed
R Garcia et al.
Applied and environmental microbiology, 51(6), 1355-1357 (1986-06-01)
Dansyl derivatives of epsilon-N-mono-, epsilon-N-di-, and epsilon-N-trimethyllysine were resolved from other amino acids in proteins by the use of high-performance liquid chromatography. The system was tested with amino acid standard combinations as well as with acid-hydrolyzed proteins known to contain
Y Minami et al.
Journal of biochemistry, 97(3), 745-753 (1985-03-01)
The amino acid sequence of a ferredoxin from a thermoacidophilic archaebacterium, Sulfolobus acidocaldarius, was determined by a combination of various conventional methods to be as follows: Gly-Ile-Asp-Pro-Tyr-Arg-Thr-His-Lys-Pro-Val-Val-Gly-Asp-Ser-Ser-Gly-His- Lys-Ile -Tyr-Gly-Pro-Val-Glu-Ser-Pro-Lys(Me)-Val-Leu-Gly-Val-His-Gly-Thr-Ile-Val -Gly-Va l-Asp-Phe-Asp-Leu-Cys-Ile-Ala-Asp-Gly-Ser-Cys-Ile-Thr-Ala-Cys-Pro-Val-As n-Val-P he-Gln-Trp-Tyr-Glu-Thr-Pro-Gly-His-Pro-Ala-Ser-Glu-Lys-Lys-Ala-Asp-Pro-V al-Asn- Glu-Gln-Ala-Cys-Ile-Phe-Cys-Met-Ala-Cys-Val-Asn-Val-Cys-Pro-Val-Ala-Ala- Ile-Asp -Val-Lys-Pro-Pro. It was composed
C G Friedrich et al.
Applied and environmental microbiology, 34(6), 706-709 (1977-12-01)
Compounds structurally related to lysine were tested against Penicillium chrysogenum Wis. 54-1255 for inhibition of growth, sporulation, and penicillin formation. This strain is relatively resistant to lysine analogs. The compounds that were the more active inhibitors of growth and whose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service