Skip to Content
Merck
All Photos(1)

Key Documents

74218

Sigma-Aldrich

H-Lys(Alloc)-OH

≥99.0%

Synonym(s):

Nδ-Alloc-L-lysine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$309.00

$309.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$309.00

About This Item

Empirical Formula (Hill Notation):
C10H18N2O4
CAS Number:
Molecular Weight:
230.26
Beilstein:
1728172
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$309.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.0%

form

solid

optical activity

[α]20/D +18.5±1°, c = 1% in 1 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

N[C@@H](CCCCNC(=O)OCC=C)C(O)=O

InChI

1S/C10H18N2O4/c1-2-7-16-10(15)12-6-4-3-5-8(11)9(13)14/h2,8H,1,3-7,11H2,(H,12,15)(H,13,14)/t8-/m0/s1

InChI key

JMZPOGGPUVRZMI-QMMMGPOBSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachel Chapla et al.
Polymers, 12(12) (2020-12-20)
Local mechanical stiffness influences cell behavior, and thus cell culture scaffolds should approximate the stiffness of the tissue type from which the cells are derived. In synthetic hydrogels, this has been difficult to achieve for very soft tissues such as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service