Skip to Content
Merck
All Photos(1)

Documents

03699

Sigma-Aldrich

Ethylenediaminetetraacetic acid tetrasodium salt hydrate

BioUltra, ≥99.0% (KT)

Synonym(s):

EDTA tetrasodium salt, Edathamil, Tetrasodium ethylenediaminetetraacetate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NaOOCCH2)2NCH2CH2N(CH2COONa)2 · xH2O
CAS Number:
Molecular Weight:
380.17 (anhydrous basis)
Beilstein:
3861753
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

product line

BioUltra

Quality Level

Assay

≥99.0% (KT)

form

powder

reaction suitability

reagent type: chelator

impurities

insoluble matter, passes filter test

pH

11.0-12.0 (25 °C, 0.1 M in H2O)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: ≤1.5
λ: 280 nm Amax: ≤0.06

SMILES string

O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.4Na.H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;;1H2/q;4*+1;/p-4

InChI key

GXFFSKMBMYHTAE-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

EDTA is an inhibitor of metalloproteases, at effective concentrations of 1-10 mM. EDTA acts as a chelator of the zinc ion in the active site of metalloproteases, and can also inhibit other metal ion-dependent proteases such as calcium-dependent cysteine proteases. For use as an anticoagulant, disodium or tripotassium salts of EDTA are most commonly used. The optimal concentration is 1.5 mg per ml of blood.

Preparation Note

Soluble in water at room temperature up to 1.45 M. The pH of this solution will be approximately 10 to 11 and will be rather viscous.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fernando Real
Experimental parasitology, 218, 107989-107989 (2020-09-18)
As the causative agent of hard-to-treat diffuse cutaneous leishmaniasis, Leishmania (L.) amazonensis persists in the host organism sheltered within large Parasitophorous Vacuoles (PVs) formed mainly in macrophages. In the present study, I present a simple and efficient method for L.
Qi-En Yang et al.
PLoS genetics, 11(7), e1005355-e1005355 (2015-07-16)
The origin of most ovarian tumors is undefined. Here, we report development of a novel mouse model in which conditional inactivation of the tumor suppressor gene Rb1 in oocytes leads to the formation of ovarian teratomas (OTs). While parthenogenetically activated
Vladimir P Baklaushev et al.
Drug delivery, 22(3), 276-285 (2014-01-21)
Targeted drug delivery for brain tumor treatment is one of the important objectives in nanomedicine. Human glioblastoma is the most frequent and aggressive type of brain tumors. The preferential expression of membrane protein connexin 43 (Cx43) and brain-specific anion transporter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service