Skip to Content
Merck
All Photos(3)

Key Documents

E26290

Sigma-Aldrich

Ethylenediaminetetraacetic acid tetrasodium salt hydrate

98%

Synonym(s):

Tetrasodium ethylenediaminetetraacetate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$41.80
500 G
$91.20
3 KG
$383.00
10 KG
$574.00

$41.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$41.80
500 G
$91.20
3 KG
$383.00
10 KG
$574.00

About This Item

Linear Formula:
(NaO2CCH2)2NCH2CH2N(CH2CO2Na)2·xH2O
CAS Number:
Molecular Weight:
380.17 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$41.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

>300 °C (lit.)

SMILES string

[Na+].[Na+].[Na+].[Na+].[H]O[H].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.4Na.H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;;1H2/q;4*+1;/p-4

InChI key

GXFFSKMBMYHTAE-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Ethylenediaminetetraacetic acid is a poylaminocarboxylic acid commonly used as a complexing agent.[1]

Application

Ethylenediaminetetraacetic acid tetrasodium salt hydrate has been used as an antimicrobial agent during biological ink preparation for use in biopolytronics.[2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An HRP-based amperometric biosensor fabricated by thermal inkjet printing.
Setti L, et al.
Sensors and Actuators B, Chemical, 126(1), 252-257 (2007)
Use of polyaminocarboxylic acids as hydrophilic masking agents for fission products in actinide partitioning processes.
Sypula M, et al.
Solvent Extr. Ion Exch., 30(7), 748-764 (2012)
Qi-En Yang et al.
PLoS genetics, 11(7), e1005355-e1005355 (2015-07-16)
The origin of most ovarian tumors is undefined. Here, we report development of a novel mouse model in which conditional inactivation of the tumor suppressor gene Rb1 in oocytes leads to the formation of ovarian teratomas (OTs). While parthenogenetically activated
Vladimir P Baklaushev et al.
Drug delivery, 22(3), 276-285 (2014-01-21)
Targeted drug delivery for brain tumor treatment is one of the important objectives in nanomedicine. Human glioblastoma is the most frequent and aggressive type of brain tumors. The preferential expression of membrane protein connexin 43 (Cx43) and brain-specific anion transporter

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service