Skip to Content
Merck
All Photos(2)

Key Documents

P2522

Sigma-Aldrich

Potassium disulfite

≥98%

Synonym(s):

Potassium metabisulfite, Potassium pyrosulfite

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$144.00

$144.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$144.00

About This Item

Linear Formula:
K2S2O5
CAS Number:
Molecular Weight:
222.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
for analytical purposes
form:
powder

$144.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

grade

for analytical purposes

Assay

≥98%

form

powder

SMILES string

[K+].[K+].[O-]S(=O)S([O-])(=O)=O

InChI

1S/2K.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

RWPGFSMJFRPDDP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium disulfite (Potassium metabisulfite, PMB) is an inorganic salt with antimicrobial properties.[1] It is a sulfiting agent that prevents browning of foods.[2] Its genotoxic and cytotoxic effect has been assessed.[1] PMB undergoes hydrolysis to form potassium bisulfite.[3]

Application

Potassium disulfite has been used in a protocol for the modification of the polydimethylsiloxane (PDMS) polymer surfaces.[4]
It may be used in the following processes:
  • Chemical etching of poly(vinylidene fluoride) in β-phase (β-PVDF) irradiated films.[5]
  • Decolorization during the synthesis of 2-iodopyrimidine derivatives.[6]
  • Palladium-catalyzed aminosulfonylation of aryl halides.[7]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biofunctionalization and self-interaction chromatography in PDMS microchannels.
Deshpande KS, et al.
Biochemical Engineering Journal, 67, 111-119 (2012)
Nanoporous β-PVDF membranes with selectively functionalized pores.
Cuscito O, et al.
Nucl. Instrum. Methods Phys. Res. Sect. B, 265(1), 309-313 (2007)
A palladium-catalyzed reaction of aryl halides, potassium metabisulfite, and hydrazines.
Ye S and Wu J.
Chemical Communications (Cambridge, England), 48(80), 10037-10039 (2012)
Amperometric quantification of sodium metabisulfite in pharmaceutical formulations utilizing tetraruthenated porphyrin film modified electrodes and batch injection analysis.
Quintino MSM, et al.
Talanta, 68(4), 1281-1286 (2006)
Gábor Vlád et al.
The Journal of organic chemistry, 67(18), 6550-6552 (2002-08-31)
A high-yield synthesis was developed for the preparation of 2,2'-bipyrimidine (1) using the Ullmann coupling of 2-iodopyrimidine. The new procedure was also used for the preparation of 4,4',6,6'-tetramethyl-2,2'-bipyrimidine (2) and 5,5'-dibromo-2,2'-bipyrimidine (3).

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service