Skip to Content
Merck
All Photos(1)

Key Documents

55777

Supelco

Potassium disulfite

analytical standard

Synonym(s):

E224, Potassium metabisulfite, Potassium pyrosulfite

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$89.20

$89.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$89.20

About This Item

Linear Formula:
K2S2O5
CAS Number:
Molecular Weight:
222.32
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$89.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98% (RT)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

H2O: 200 mg/mL, clear to very slightly hazy, colorless

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[K+].[K+].[O-]S(=O)S([O-])(=O)=O

InChI

1S/2K.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

RWPGFSMJFRPDDP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium disulfite (Potassium metabisulfite, PMB) is an inorganic salt with antimicrobial properties. It is a sulfiting agent that prevents browning of foods.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sven Hildebrand et al.
Scientific reports, 9(1), 10880-10880 (2019-07-28)
Optical clearing techniques and light sheet microscopy have transformed fluorescent imaging of rodent brains, and have provided a crucial alternative to traditional confocal or bright field techniques for thin sections. However, clearing and labeling human brain tissue through all cortical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service