Skip to Content
Merck
All Photos(1)

Key Documents

71970

Sigma-Aldrich

Sodium sulfate decahydrate

puriss. p.a., crystallized, ≥99.0% (calc. based on dry substance, T)

Synonym(s):

Glauber’s salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$103.00

$103.00


Estimated to ship onApril 03, 2025


Request a Bulk Order

Select a Size

Change View
1 KG
$103.00

About This Item

Linear Formula:
Na2SO4 · 10H2O
CAS Number:
Molecular Weight:
322.20
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (calc. based on dry substance, T)
form:
crystals

$103.00


Estimated to ship onApril 03, 2025


Request a Bulk Order

grade

puriss. p.a.

Quality Level

Assay

≥99.0% (calc. based on dry substance, T)

form

crystals

quality

crystallized

impurities

≤0.0005% total nitrogen (N)

loss

53-57% loss on drying, 130 °C

pH

5.0-8.0 (25 °C, 50 mg/mL in H2O)

density

1.46 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤10 mg/kg

cation traces

Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤20 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S.10H2O/c;;1-5(2,3)4;;;;;;;;;;/h;;(H2,1,2,3,4);10*1H2/q2*+1;;;;;;;;;;;/p-2

InChI key

RSIJVJUOQBWMIM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium sulfate decahydrate (Glauber′s salt, mirabilite, Na2SO4·10H2O) is the decahydrate salt of sodium sulfate. Its crystal structure has been investigated by single-crystal neutron diffraction studies.[1] Its crystallization enthalpy has been evaluated.[2] It can be synthesized by reacting MnSO4, thiophene-2,5-dicarboxylic acid and sodium glutamate.[3]

Application


  • Nanoscale Stabilization Mechanism of Sodium Sulfate Decahydrate at Polyelectrolyte Interfaces: Discusses the stabilization mechanisms at the nanoscale, providing insights into sodium sulfate decahydrate′s applications in polyelectrolyte-enhanced processes, relevant in materials science and engineering (Akamo et al., 2024).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrothermal Synthesis and Crystal Structure of A Special Sodium Sulfate Decahydrate (Na2SO4?10H2O).
Zhang L and He YX.
Asian Journal of Chemistry, 26(12) (2014)
Crystal structures of sodium sulfate decahydrate (Glauber's salt) and sodium tetraborate decahydrate (borax). Redetermination by neutron diffraction.
Levy HA and Lisensky GC.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 34(12), 3502-3510 (1978)
Thermodynamics of crystallization of sodium sulfate decahydrate in H2O-NaCl-Na2SO4: application to Na2SO4?10H2O-based latent heat storage materials.
Marliacy P, et al.
Thermochimica Acta, 344(1), 85-94 (2000)
Jia-ying Zhu et al.
Zhongguo wei zhong bing ji jiu yi xue = Chinese critical care medicine = Zhongguo weizhongbing jijiuyixue, 24(6), 346-348 (2012-10-02)
To investigate the effect of nasal tube feeding of rhubarb solution and umbilical compress of Glauber's salt for purgation in the treatment of patients with severe acute organophosphorus pesticide poisoning (AOPP). A retrospective study was conducted. Eighty patients with severe
Shijie Liu et al.
The Journal of biological chemistry, 288(13), 8794-8803 (2013-02-09)
Lgr4/Gpr48 is one of the newly identified R-spondins receptors and potentiates Wnt signaling, which regulates intestinal homeostasis. We used a hypomorphic mouse strain to determine the role of Lgr4 in intestinal inflammation and recovery. Intestinal inflammation was induced with dextran

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service