Skip to Content
Merck
All Photos(1)

Key Documents

246980

Sigma-Aldrich

Sodium sulfate decahydrate

reagent grade, 97%

Synonym(s):

Glauber’s salt

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$121.00

$121.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
$121.00

About This Item

Linear Formula:
Na2SO4 · 10H2O
CAS Number:
Molecular Weight:
322.20
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent grade
form:
crystals or granules

$121.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

reagent grade

Assay

97%

form

crystals or granules

pH

5-7

density

1.46 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S.10H2O/c;;1-5(2,3)4;;;;;;;;;;/h;;(H2,1,2,3,4);10*1H2/q2*+1;;;;;;;;;;;/p-2

InChI key

RSIJVJUOQBWMIM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrothermal Synthesis and Crystal Structure of A Special Sodium Sulfate Decahydrate (Na2SO4?10H2O).
Zhang L and He YX.
Asian Journal of Chemistry, 26(12) (2014)
Crystal structures of sodium sulfate decahydrate (Glauber's salt) and sodium tetraborate decahydrate (borax). Redetermination by neutron diffraction.
Levy HA and Lisensky GC.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 34(12), 3502-3510 (1978)
Thermodynamics of crystallization of sodium sulfate decahydrate in H2O-NaCl-Na2SO4: application to Na2SO4?10H2O-based latent heat storage materials.
Marliacy P, et al.
Thermochimica Acta, 344(1), 85-94 (2000)
C Poelaert et al.
Journal of animal science, 90 Suppl 4, 387-389 (2013-02-13)
Over the past decade, several in vitro methods have been developed to study intestinal fermentation in pigs and its influence on health. In these methods, samples are fermented by a bacterial inoculum diluted in a mineral buffer solution. Generally, a
Clément Cartier et al.
Water research, 47(2), 661-671 (2012-11-24)
Release of lead from 80% partially replaced service lines was compared to full lead service lines using harvested-stabilized lead pipes and field brass connectors. After more than a year of stabilization, lead release was consistent with field samples. Over the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service