Skip to Content
Merck
All Photos(1)

Key Documents

71382

Sigma-Aldrich

Sodium chloride

puriss. p.a., ≥99.5% (AT), powder or crystals

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
powder or crystals
solubility:
water: soluble 358 g/L at 20 °C
Pricing and availability is not currently available.

vapor pressure

1.33 hPa ( 865 °C)

grade

puriss. p.a.

Assay

≥99.5% (AT)

form

powder or crystals

impurities

Traces of [Fe(CN)6]4- (information only)
≤0.001% total nitrogen (N)

pH

5.0-8.0 (25 °C, 50 mg/mL in H2O)

mp

801 °C (lit.)

solubility

water: soluble 358 g/L at 20 °C

anion traces

bromide (Br-): ≤50 mg/kg
iodide (I-): ≤10 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium chloride is an ionic compound that comprises 1:1 ratio of sodium and chloride ions. It is also known as table salt, common salt, or halite. It is used as a raw material for the production of many chemicals which include, sodium carbonate, sodium hydrogen carbonate, and sodium dihydrogen phosphate.

Application


  • Sodium alginate edible coating containing Ferulago angulata (Schlecht.) Boiss essential oil, nisin, and NaCl: Its impact on microbial, chemical, and sensorial properties of refrigerated chicken breast.: This study investigates the use of sodium chloride in sodium alginate edible coatings, focusing on its effects on the microbial, chemical, and sensorial properties of refrigerated chicken breast (Panahi & Mohsenzadeh, 2022).

  • Physicochemical stability of PF-06439535 (bevacizumab-bvzr; Zirabev), a bevacizumab biosimilar, under extended in-use conditions.: The research investigates the physicochemical stability of a bevacizumab biosimilar, highlighting the role of sodium chloride in maintaining stability under extended in-use conditions (Weiser et al., 2023).


Other Notes

Sales restrictions may apply

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriel L Converse et al.
Journal of the mechanical behavior of biomedical materials, 2(6), 627-635 (2009-09-01)
The apparent mechanical properties of hydroxyapatite (HA) whisker reinforced polyetherketoneketone (PEKK) scaffolds were evaluated in unconfined, uniaxial compression to investigate the effects of the porosity (75%, 82.5% and 90%), HA content (0, 20 and 40 vol%) and mold temperature (350
Timothy L Conrad et al.
Journal of the mechanical behavior of biomedical materials, 106, 103730-103730 (2020-04-07)
Porous polyetheretherketone (PEEK) and bioactive hydroxyapatite (HA) reinforced PEEK scaffolds have attracted recent interest for enabling biologic fixation of orthopaedic and spinal implants, such as interbody spinal fusion cages. Porous PEEK and HA-PEEK scaffolds have been prepared by compression molding
Rebecca Soffe et al.
PloS one, 14(6), e0218102-e0218102 (2019-06-07)
Artificial surfaces are routinely used instead of leaves to enable a reductionist approach in phyllosphere microbiology, the study of microorganisms residing on plant leaf surfaces. Commonly used artificial surfaces include, flat surfaces, such as metal and nutrient agar, and microstructured
A M Gibson et al.
International journal of food microbiology, 6(2), 155-178 (1988-03-01)
The growth responses of salmonellae (mixed inoculum of Salmonella thompson, S. stanley and S. infantis) as affected by NaCl concentration, pH level and storage temperature were studied in laboratory medium. Growth curves were obtained at 5 concentrations of NaCl (0.5-4.5%
Akio Suzuki et al.
PloS one, 14(6), e0217944-e0217944 (2019-06-07)
Anti-prion protein (PrP) monoclonal antibody 132, which recognizes mouse PrP amino acids 119-127, enables us to reliably detect abnormal isoform prion protein (PrPSc) in cells or frozen tissue sections by immunofluorescence assay, although treatment with guanidinium salts is a prerequisite.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service