Skip to Content
Merck
All Photos(1)

Documents

63062

Sigma-Aldrich

Magnesium carbonate basic

purum p.a., heavy, ≥40% (MgO)

Synonym(s):

Magnesium hydroxide carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCO3
CAS Number:
Molecular Weight:
84.31
EC Number:
MDL number:
UNSPSC Code:
51171508
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

form

powder

concentration

≥40% (MgO)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤6000 mg/kg

cation traces

Ca: ≤1000 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤2000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Mg++].O[Mg]O.[O-]C([O-])=O

InChI

1S/CH2O3.2Mg.2H2O/c2-1(3)4;;;;/h(H2,2,3,4);;;2*1H2/q;2*+2;;/p-4

InChI key

UDYRPJABTMRVCX-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium carbonate basic (Magnesium hydroxy carbonate) is a magnesium salt. It has been synthesized by reacting magnesium hydroxide and carbon dioxide.

Application

Magnesium carbonate basic may be employed for the isolation of carotenoids from grapes during the quantification of carotenoids in grapes by reversed-phase C30 (RP-30) HPLC-DAD-MS (ESI+) (High-performance liquid chromatography-diode array detection-mass spectrometry with electrospray ionization) analysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of a magnesium hydroxy carbonate from magnesium hydroxide.
Botha A and Strydom CA.
Hydrometallurgy, 62(1), 175-183 (2001)
Pasquale Crupi et al.
Journal of mass spectrometry : JMS, 45(9), 971-980 (2010-09-04)
Carotenoids are uniquely functional polyene pigments ubiquitous in nature; aside from being responsible for the color of a wide variety of vegetables, interest is being focused on food carotenoids due to their likely health benefits. From analytical point of view

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service