Skip to Content
Merck
All Photos(4)

Key Documents

239267

Sigma-Aldrich

Cobalt(II) nitrate hexahydrate

ACS reagent, ≥98%

Synonym(s):

Cobaltous nitrate, Cobaltous nitrate hexahydrate, Nitric acid, cobalt(II) salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$158.00
100 G
$239.00
500 G
$906.00

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$158.00
100 G
$239.00
500 G
$906.00

About This Item

Linear Formula:
Co(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
291.03
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
chips
powder, crystals or chunks
solid

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

ACS reagent

Quality Level

Assay

≥98%
98.0-102.0%

form

chips
powder, crystals or chunks
solid

reaction suitability

reagent type: catalyst
core: cobalt

impurities

≤0.01% insolubles

pH

4 (20 °C, 100 g/L)

mp

55 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Cu: ≤0.002%
Fe: ≤0.001%
K: ≤0.01%
Mg: ≤0.005%
NH4+: ≤0.2%
Na: ≤0.05%
Ni: ≤0.15%
Pb: ≤0.002%
Zn: ≤0.01%

SMILES string

O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Co.2NO3.6H2O/c;2*2-1(3)4;;;;;;/h;;;6*1H2/q+2;2*-1;;;;;;

InChI key

QGUAJWGNOXCYJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cobalt(II)nitrate hexahydrate is a red-brown crystalline structure that is soluble in water and organic solvents. It is commonly used in the preparation of cobalt-based catalysts, pigments, chemicals, and vitamin B12 preparations. Additionally, it serves high-purity cobalt source for use in the electronics industry. [1] [2]

Application

Cobalt(II)nitrate hexahydrate can be used as a catalyst to synthesize:
  • 5-Carboxanilide-dihydropyrimidinone derivatives by the condensation of acetoacetanilides, aldehydes, and urea/thiourea.
  • Nitro indoles by the nitration of indoles using tert-butyl nitrite as the nitro source.
  • Quinazolinones from 2-aminobenzonitriles using alcohols in the presence of phosphine ligand.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of a cobalt metal-organic framework (Co-MOF) and its application as a polypropylene flame retardant by compounding with melamine polyphosphate
Ya Xu, et al.
Polymer Testing, 116, 107765-107765 (2022)
Chlorinated drinking water; chlorination byproducts; some other halogenated compounds; cobalt and cobalt compounds
International Agency for Research on Cancer and others
IARC Monographs on the Evaluation of Carcinogenic Risks to Humans / World Health Organization, International Agency for Research on Cancer, 52, 363-472 (1991)
Some Phosphine Oxide and Arsine Oxide Complexes of Cobalt (II) Nitrate.
Cotton FA, et al.
Inorganic Chemistry, 2(6), 1162-1165 (1963)
Horacio López-Sandoval et al.
Journal of inorganic biochemistry, 102(5-6), 1267-1276 (2008-02-29)
In this work we present the synthesis, structural and spectroscopic characterisation of a series of cobalt(II) and zinc(II) coordination compounds with benzimidazole (bz) and its 2-benzimidazole derivatives: 2-aminobenzimidazole (2ab), albendazole (abz) and tris(2-benzimidazolylmethyl)amine (ntb). The compounds were evaluated for their
Thermal decomposition of cobalt nitrato compounds: Preparation of anhydrous cobalt (II) nitrate and its characterisation by Infrared and Raman spectra.
Ehrhardt C, et al.
Thermochimica Acta, 432(1), 36-40 (2005)

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. 239267와 관련하여 입자크기를 알 수 있을까요? 저희가 원하는 사이즈는 <10μm 이하의 파우더를 구입하고 싶습니다 해당되는 스펙의 파우더가 있다면 알려주시기 바랍니다

    1 answer
    1. Particle size testing is not performed on this product and there are no options for cobalt(II) nitrate hexahydrate with particle size testing. Please consider grinding to the appropriate particle size.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service