Skip to Content
Merck
All Photos(3)

Key Documents

60489

Sigma-Aldrich

Potassium peroxodisulfate

puriss. p.a., ACS reagent, ≥99.0% (RT)

Synonym(s):

Potassium persulfate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$125.00
1 KG
$335.00

$125.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 11, 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
$125.00
1 KG
$335.00

About This Item

Linear Formula:
K2S2O8
CAS Number:
Molecular Weight:
270.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
powder or crystals

$125.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 11, 2025Details


Request a Bulk Order

grade

ACS reagent
puriss. p.a.

Assay

≥99.0% (RT)

form

powder or crystals

impurities

≤0.001% total nitrogen (N)

pH

2.5-4.5 (25 °C, 27 g/L)

anion traces

chloride (Cl-): ≤10 mg/kg

cation traces

Ag: ≤5 mg/kg
Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Li: ≤5 mg/kg
Mg: ≤100 mg/kg
Mn: ≤1 mg/kg
Mo: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Tl: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

forensics and toxicology

SMILES string

[K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O

InChI

1S/2K.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h;;(H,1,2,3)(H,4,5,6)/q2*+1;/p-2

InChI key

USHAGKDGDHPEEY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium peroxodisulfate is also known as potassium persulfate (K2S2O8). It is commonly used as an oxidizing agent and polymerization initiator in organic synthesis.

Application

Potassium peroxodisulfate can be used as:      
  • A polymerization initiator in the synthesis of polyacrylamide via radical polymerization of acrylamide.      
  • An oxidizing agent in the photochemical oxidation of atrazine aqueous solution.

K2S2O8 can also be used in the following process:      
  • K2S2O8-AgSCF3 catalytic system is used in the direct trifluoromethylthiolation of unactivated C(sp3)-H bonds under mild conditions. Usage of K2S2O8 in this process plays an important role in the activation of the C-H bond and the oxidation of AgSCF3.
  • K2S2O8-FeCl3 is used for the thiocyanation of 2H-indazoles using ammonium thiocyanate as a thiocyanating agent.

Other Notes

Oxidizing agent for decomposition by fusion[1][2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service