Skip to Content
Merck
All Photos(1)

Key Documents

14372

Sigma-Aldrich

Pumice stone

granulated

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
NA.47

form

granular

quality

granulated

particle size

2-5 mm (determined by sieving)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pitsapaporn Wangpermtam et al.
The journal of adhesive dentistry, 13(5), 445-453 (2011-09-22)
To determine the adhesion of resin-modified glass-ionomer cement to bovine dentin under contaminated and decontaminated conditions. Forty-five bovine mandibular incisors were used. The surfaces of bovine dentin specimens were subjected to Temp-bond, dental handpiece lubricant (contamination), Hibiscrub, chlorhexidine or pumice
A Sohrabi et al.
European journal of paediatric dentistry : official journal of European Academy of Paediatric Dentistry, 13(4), 317-320 (2012-12-29)
Self-etch bonding systems have been introduced recently and have the advantage of reduced operating time. Because of patient management and isolation difficulties in children, the present study aimed to evaluate microtensile bond strength of two self-etch bonding systems (AdheSE and
Usama Beshay et al.
Polish journal of microbiology, 60(2), 133-138 (2011-09-13)
The studies have been performed to analyze the production of beta-glucanase by a recombinant strain of Escherichia coli immobilized in different matrices. Porous sintered glass SIRAN, Ceramic supporting matrices and Broken Pumice stone as well as SIRAN Raschig-rings were examined
Penicillium digitatum immobilized on pumice stone as a new solid phase extractor for preconcentration and/or separation of trace metals in environmental samples.
Baytak S, et al.
Journal of Hazardous Materials, 153(3), 975-983 (2008)
Martin D Brasier et al.
Astrobiology, 11(7), 725-735 (2011-09-02)
The context for the emergence of life on Earth sometime prior to 3.5 billion years ago is almost as big a puzzle as the definition of life itself. Hitherto, the problem has largely been addressed in terms of theoretical and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service