Skip to Content
Merck
All Photos(1)

Key Documents

PX1980

Supelco

Pumice Stone

size <400 mesh

Synonym(s):

Pumice Stone, Pumice

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11111610
Pricing and availability is not currently available.

grade

for elemental analysis

Quality Level

form

powder

size

<400 mesh

pH

7.5- 8.0

shipped in

ambient

storage temp.

room temp

SMILES string

CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)F)O)C)C)OC(=O)CC

InChI

1S/C28H37FO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-21,31H,6-9,12,14-15H2,1-5H3

InChI key

CIWBQSYVNNPZIQ-UHFFFAOYSA-N

General description

< 400 Mesh

Application

<400 Mesh

Analysis Note

Description: Fine gray powder
Mesh size (<140): 75% min

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service