Skip to Content
Merck
All Photos(1)

Documents

Z802220

Specac® Quest ATR diamond accessory

Color Black

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.32

manufacturer/tradename

Specac Ltd

technique(s)

IR spectroscopy: suitable

General description

The Quest ATR Accessory is a performance single-reflection ATR accessory from Specac designed for laboratory spectroscopic sample analysis in the mid- and far-infrared. With innovative optical design and durable monolithic diamond ATR crystal option, this sets the benchmark in performance and value for ATR spectroscopy.
The Quest′s innovative optical design is based around all reflective gold-coated optics, and features Specac′s Synopti-Focal Array technology with precision-moulded aspheric mirrors. These features allow for a high throughput accessory with an extended wavelength capability suitable for both mid and far-IR spectroscopy.
The following ATR crystal puck options are currently available:
  • High throughput diamond (Z802220)
  • Extended wavelength diamond (Z802344)

Each puck is made from hardened stainless steel and are held in place on the accessory magnetically making them easily interchangeable without requiring additional alignment.
Requires a Specac Benchmark baseplate suitable for your IR spectrometer, which is not included and needs to be ordered separately unless already available in your lab.

Legal Information

Specac is a registered trademark of Specac Ltd.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service