MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
Skip To
Z802344
Specac®Quest ATR diamond extended wavelength accessory
Color Black
Sign In to View Organizational & Contract Pricing.
About This Item
UNSPSC Code:
12352200
General description
The Quest ATR Accessory is a performance single-reflection ATR accessory from Specac designed for laboratory spectroscopic sample analysis in the mid- and far-infrared. With innovative optical design and durable monolithic diamond ATR crystal option, this sets the benchmark in performance and value for ATR spectroscopy.
The Quest′s innovative optical design is based around all reflective gold-coated optics, and features Specac′s Synopti-Focal Array technology with precision-moulded aspheric mirrors. These features allow for a high throughput accessory with an extended wavelength capability suitable for both mid and far-IR spectroscopy.
The following ATR crystal puck options are currently available:
  • High throughput diamond (Z802220)
  • Extended wavelength diamond (Z802344)
Each puck is made from hardened stainless steel and are held in place on the accessory magnetically making them easily interchangeable without requiring additional alignment.
Requires a Specac Benchmark baseplate suitable for your IR spectrometer, which is not included and needs to be ordered separately unless already available in your lab.
Legal Information
Specac is a registered trademark of Specac Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)