Skip to Content
Merck
All Photos(1)

Key Documents

Y0001334

Clopidogrel for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(S)-Methyl (2-chlorophenyl)(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate hydrogensulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16ClNO2S · H2SO4
CAS Number:
Molecular Weight:
419.90
Beilstein:
9967887
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

clopidogrel

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.COC(=O)[C@@H](N1CCc2sccc2C1)c3ccccc3Cl

InChI

1S/C16H16ClNO2S.H2O4S/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-14-11(10-18)7-9-21-14;1-5(2,3)4/h2-5,7,9,15H,6,8,10H2,1H3;(H2,1,2,3,4)/t15-;/m0./s1

InChI key

FDEODCTUSIWGLK-RSAXXLAASA-N

Gene Information

human ... P2RY12(64805)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clopidogrel for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinmeng Chen et al.
Biopharmaceutics & drug disposition, 36(1), 64-68 (2014-09-27)
Erigeron breviscapus (Vant.) Hand-Mazz, a traditional Chinese medicine, is often co-prescribed with clopidogrel for the treatment of ischemic vascular diseases. Scutellarin is the representative bioactive flavonoid isolated from this herb. The aim of this study was to explore the effect
Kyoung-Woo Seo et al.
Clinical therapeutics, 36(11), 1588-1594 (2014-09-15)
Platelet-function suppression with antiplatelet therapy is effective in preventing and treating cardiovascular disease. Clopidogrel is a thienopyridine derivative that blocks platelet activation by adenosine diphosphate receptor binding. This study demonstrates the effects of generic clopidogrel bisulfate in comparison to branded
Matteo Tozzi et al.
Journal of vascular surgery, 60(4), 1019-1023 (2014-05-14)
An alternative surgical technique for the repair an aneurysmatic arteriovenous fistula (AVF) using a staple remodeling operation has been developed and the results are reported. All patients presenting with an aneurysmatic autogenous AVF of the upper extremities between January 2012
Lucia Stefanini et al.
The Journal of clinical investigation, 125(4), 1419-1432 (2015-02-24)
The small GTPase RAP1 is critical for platelet activation and thrombus formation. RAP1 activity in platelets is controlled by the GEF CalDAG-GEFI and an unknown regulator that operates downstream of the adenosine diphosphate (ADP) receptor, P2Y12, a target of antithrombotic
Wei Li et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 88(3), 706-717 (2014-10-14)
Chemical modification of nanoclay will ensure further progress on these materials. In this work, we show that montmorillonite (MTM) nanosheets can be modified with β-cyclodextrin (CD) via a nucleophilic substitution reaction between mono-6-(p-toluenesulfonyl)-6-deoxy-β-CD and an amino group of 3-aminopropyltriethoxysilane (APTES)-functionalized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service