Skip to Content
Merck
All Photos(1)

Key Documents

C0614

Sigma-Aldrich

(±) Clopidogrel hydrogensulfate

≥98% (HPLC), powder

Synonym(s):

Methyl (2-chlorophenyl)(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate hydrogen sulfate, SR-25990

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$327.00
25 MG
$1,260.00

$327.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$327.00
25 MG
$1,260.00

About This Item

Empirical Formula (Hill Notation):
C16H16ClNO2S · H2SO4
CAS Number:
Molecular Weight:
419.90
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$327.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white

solubility

DMSO: ~26 mg/mL

originator

Bristol-Myers Squibb

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.COC(=O)C(N1CCc2sccc2C1)c3ccccc3Cl

InChI

1S/C16H16ClNO2S.H2O4S/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-14-11(10-18)7-9-21-14;1-5(2,3)4/h2-5,7,9,15H,6,8,10H2,1H3;(H2,1,2,3,4)

InChI key

FDEODCTUSIWGLK-UHFFFAOYSA-N

Biochem/physiol Actions

Inhibits ADP-induced platelet aggregation; anti-thrombotic drug.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the P2 Receptors: P2Y G-Protein Family page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Bristol-Myers Squibb. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Packaging

Store under desiccant

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Wang et al.
Nature protocols, 6(7), 935-942 (2011-07-02)
We describe an effective and simple protocol that uses click chemistry to attach native β-cyclodextrin (β-CD) to silica particles, resulting in a chiral stationary phase (CCNCSP) that can be used for the enantioseparation of chiral drugs by high-performance liquid chromatography

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Discover Bioactive Small Molecules for ADME/Tox

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service