Skip to Content
Merck
All Photos(1)

Key Documents

Y0001300

Sodium cromoglicate for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cromolyn sodium salt, Cromoglycate, Cromoglycic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C23H14O11Na2
CAS Number:
Molecular Weight:
512.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

sodium cromoglicate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na].OC(COc1cccc2OC(=CC(=O)c12)C(O)=O)COc3cccc4OC(=CC(=O)c34)C(O)=O

InChI

1S/C23H16O11.Na.H/c24-11(9-31-14-3-1-5-16-20(14)12(25)7-18(33-16)22(27)28)10-32-15-4-2-6-17-21(15)13(26)8-19(34-17)23(29)30;;/h1-8,11,24H,9-10H2,(H,27,28)(H,29,30);;

InChI key

UXMUBZWVLQEMRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium cromoglicate for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Cromolyn blocks the release of histamine and other pro-inflammatory mediators from mast cells.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eyal Zur
International journal of pharmaceutical compounding, 16(5), 386-393 (2012-10-18)
Sodium cromoglicate (cromolyn sodium) is a very well-known medicine that has been used for many years for various allergic conditions. The topical use of this medicine is less known, and there are no commercial medicines of cream, gel, or lotion
Lindsey L Kennedy et al.
Laboratory investigation; a journal of technical methods and pathology, 94(12), 1406-1418 (2014-11-05)
Cholangiopathies are characterized by dysregulation of the balance between biliary growth and loss. We have shown that histamine (HA) stimulates biliary growth via autocrine mechanisms. To evaluate the paracrine effects of mast cell (MC) stabilization on biliary proliferation, sham or
Fuat Bicer et al.
American journal of physiology. Renal physiology, 308(2), F103-F113 (2014-09-12)
The cause of chronic pelvic pain in interstitial cystitis/painful bladder syndrome (IC/PBS) remains unclear; autoimmunity is a possible etiology. We have recently shown that injection of a single immunogenic peptide of uroplakin 3A (UPK3A 65-84) induces experimental autoimmune cystitis (EAC)
Yuan-yuan Ma et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 63(7), 527-537 (2014-03-08)
Mast cell (MC) degranulation can break peripheral immune tolerance. However, its mechanism remains unclear. Our goal was to study the stabilization of MC membranes by heme oxygenase-1 (HO-1) in order to influence dendritic cell (DC) function. Mast cells and dendritic
Craig M McKittrick et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 35(4), 638-647 (2015-01-08)
Blood brain barrier (BBB) breakdown and neuroinflammation are key events in ischemic stroke morbidity and mortality. The present study investigated the effects of mast cell deficiency and stabilization on BBB breakdown and neutrophil infiltration in mice after transient middle cerebral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service