Merck
  • Inhibiting mast cell degranulation by HO-1 affects dendritic cell maturation in vitro.
Inhibiting mast cell degranulation by HO-1 affects dendritic cell maturation in vitro.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2014-03-08)
Yuan-yuan Ma, Mu-qing Yang, Chun-feng Wang, Jing Ding, Ji-yu Li
ABSTRACT
Mast cell (MC) degranulation can break peripheral immune tolerance. However, its mechanism remains unclear. Our goal was to study the stabilization of MC membranes by heme oxygenase-1 (HO-1) in order to influence dendritic cell (DC) function. Mast cells and dendritic cells were prepared from 8-week-old to 10-week-old C57BL/6 mice; spleen mononuclear cells (SMCs) were prepared from 8-week-old to 10-week-old C57BL/6 and Balb/c mice. Mast cells were pretreated with PBS, DMSO, Hemin (50 μl/ml), and Znpp (50 μl/ml) for 8 h. Real-time PCR and western-blot tested the HO-1 of MC mRNA and protein. The co-stimulatory molecules of DCs (CD80, CD86, CD40) were measured by flow cytometry, and levels of TNF-α, IL-6, and IFN-γ were measured by ELISA. We set up a one-way mixed lymphocyte reaction (MLR) model to test the proliferation of SMCs after MC/DC interaction. *P < 0.05 (t test) was taken as the level of statistical significance. MCs pretreated with hemin induced HO-1 mRNA and protein expression, then interacted with DCs; expression of the co-stimulatory molecules was attenuated. The TNF-α, IL-6, and IFN-γ levels in the co-culture system were decreased. These DCs couldn't stimulate the proliferation of SMCs. Inhibiting MC degranulation by HO-1 restrained DC maturation and attenuated the proliferation of SMCs.
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Cromolyn sodium salt, ≥95%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sodium cromoglicate for system suitability, European Pharmacopoeia (EP) Reference Standard